gRNA Design Details
- Target DNA Sequence
- TAACCAATCCGAAAAGAGCG
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 49742247 - 49742269 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
97.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMBIM6
- Entrez Gene ID
-
7009
- Gene Name
- transmembrane BAX inhibitor motif containing 6
- Gene Aliases
-
BAXI1, BI-1, TEGT
- Chromosome Location
-
Chr.12: 49741510 - 49764934 on GRCh38
- UniGene ID
- Hs.743965
- Species
-
Human