gRNA Design Details

Target DNA Sequence
CCGAACCAAGTAGGAAACCC
PAM Sequence
CGG
Target locus
Chr.1: 154970032 - 154970054 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 91.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SHC1
Entrez Gene ID
6464
Gene Name
SHC adaptor protein 1
Gene Aliases
SHC, SHCA
Chromosome Location
Chr.1: 154962298 - 154974492 on GRCh38
UniGene ID
Hs.433795
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001130040.1 NP_001123512.1 1
NM_001130041.1 NP_001123513.1 2
NM_001202859.1 NP_001189788.1 2
NM_003029.4 NP_003020.2 2
NM_183001.4 NP_892113.4 1
XM_005245449.4 XP_005245506.1 1
XM_005245451.4 XP_005245508.1 2
XM_011509892.2 XP_011508194.1 1
XM_011509893.2 XP_011508195.1 1
XM_011509894.2 XP_011508196.1 1
XM_011509897.1 XP_011508199.1 2
XM_017002081.1 XP_016857570.1 1
XM_017002082.1 XP_016857571.1 1
XM_017002083.1 XP_016857572.1 2