Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001130040.1 | NP_001123512.1 | 1 |
NM_001130041.1 | NP_001123513.1 | 2 | |
NM_001202859.1 | NP_001189788.1 | 2 | |
NM_003029.4 | NP_003020.2 | 2 | |
NM_183001.4 | NP_892113.4 | 1 | |
XM_005245449.4 | XP_005245506.1 | 1 | |
XM_005245451.4 | XP_005245508.1 | 2 | |
XM_011509892.2 | XP_011508194.1 | 1 | |
XM_011509893.2 | XP_011508195.1 | 1 | |
XM_011509894.2 | XP_011508196.1 | 1 | |
XM_011509897.1 | XP_011508199.1 | 2 | |
XM_017002081.1 | XP_016857570.1 | 1 | |
XM_017002082.1 | XP_016857571.1 | 1 | |
XM_017002083.1 | XP_016857572.1 | 2 |