gRNA Design Details
- Target DNA Sequence
- GTCCTTAAACCGCCCACTCA
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 118262595 - 118262617 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
95.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MPZL2
- Entrez Gene ID
-
10205
- Gene Name
- myelin protein zero like 2
- Gene Aliases
-
EVA, EVA1
- Chromosome Location
-
Chr.11: 118253416 - 118264536 on GRCh38
- UniGene ID
- Hs.116651
- Species
-
Human