gRNA Design Details
- Target DNA Sequence
- GACCTCAGCACCGCTAACAG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 209653212 - 209653234 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
93.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MAP2
- Entrez Gene ID
-
4133
- Gene Name
- microtubule associated protein 2
- Gene Aliases
-
MAP2A, MAP2B, MAP2C
- Chromosome Location
-
Chr.2: 209424047 - 209734118 on GRCh38
- UniGene ID
- Hs.368281
- Species
-
Human