gRNA Design Details
- Target DNA Sequence
- GGAGAAAGACAAGAAGGCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 17275276 - 17275298 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
76.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BASP1
- Entrez Gene ID
-
10409
- Gene Name
- brain abundant membrane attached signal protein 1
- Gene Aliases
-
CAP-23, CAP23, NAP-22, NAP22
- Chromosome Location
-
Chr.5: 17216823 - 17276845 on GRCh38
- UniGene ID
- Hs.201641
- Species
-
Human