gRNA Design Details
- Target DNA Sequence
- ACTGTAGTATCAGTCAGTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 19378478 - 19378500 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
75.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RPS6
- Entrez Gene ID
-
6194
- Gene Name
- ribosomal protein S6
- Gene Aliases
-
S6
- Chromosome Location
-
Chr.9: 19376256 - 19380237 on GRCh38
- UniGene ID
- Hs.408073
- Species
-
Human