gRNA Design Details
- Target DNA Sequence
- CAAGGTTTCCCCATGAAGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 19378860 - 19378882 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
63.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RPS6
- Entrez Gene ID
-
6194
- Gene Name
- ribosomal protein S6
- Gene Aliases
-
S6
- Chromosome Location
-
Chr.9: 19376256 - 19380237 on GRCh38
- UniGene ID
- Hs.408073
- Species
-
Human