gRNA Design Details

Target DNA Sequence
ATGAGGGACTTTGCTAACCA
PAM Sequence
GGG
Target locus
Chr.14: 33215348 - 33215370 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 87.81/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
NPAS3
Entrez Gene ID
64067
Gene Name
neuronal PAS domain protein 3
Gene Aliases
MOP6, PASD6, bHLHe12
Chromosome Location
Chr.14: 32934785 - 33804176 on GRCh38
UniGene ID
Hs.657892
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001164749.1 NP_001158221.1 3
NM_001165893.1 NP_001159365.1 2
NM_022123.2 NP_071406.1 2
NM_173159.2 NP_775182.1 2
XM_005267991.3 XP_005268048.2 3
XM_005267992.3 XP_005268049.2 3
XM_011537067.2 XP_011535369.1 3
XM_011537069.2 XP_011535371.2 3
XM_011537070.2 XP_011535372.1 2
XM_011537071.2 XP_011535373.2 2
XM_011537072.2 XP_011535374.1 2
XM_017021582.1 XP_016877071.1 3
XM_017021583.1 XP_016877072.1 3
XM_017021584.1 XP_016877073.1 2
XM_017021585.1 XP_016877074.1 2
XM_017021586.1 XP_016877075.1 2
XM_017021587.1 XP_016877076.1 2
XM_017021588.1 XP_016877077.1 3