Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001164749.1 | NP_001158221.1 | 3 |
NM_001165893.1 | NP_001159365.1 | 2 | |
NM_022123.2 | NP_071406.1 | 2 | |
NM_173159.2 | NP_775182.1 | 2 | |
XM_005267991.3 | XP_005268048.2 | 3 | |
XM_005267992.3 | XP_005268049.2 | 3 | |
XM_011537067.2 | XP_011535369.1 | 3 | |
XM_011537069.2 | XP_011535371.2 | 3 | |
XM_011537070.2 | XP_011535372.1 | 2 | |
XM_011537071.2 | XP_011535373.2 | 2 | |
XM_011537072.2 | XP_011535374.1 | 2 | |
XM_017021582.1 | XP_016877071.1 | 3 | |
XM_017021583.1 | XP_016877072.1 | 3 | |
XM_017021584.1 | XP_016877073.1 | 2 | |
XM_017021585.1 | XP_016877074.1 | 2 | |
XM_017021586.1 | XP_016877075.1 | 2 | |
XM_017021587.1 | XP_016877076.1 | 2 | |
XM_017021588.1 | XP_016877077.1 | 3 |