gRNA Design Details
- Target DNA Sequence
- GGTGATAATGACAGCCACCA
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 1754440 - 1754462 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADIPOR2
- Entrez Gene ID
-
79602
- Gene Name
- adiponectin receptor 2
- Gene Aliases
-
ACDCR2, PAQR2
- Chromosome Location
-
Chr.12: 1690981 - 1788679 on GRCh38
- UniGene ID
- Hs.371642
- Species
-
Human