gRNA Design Details
- Target DNA Sequence
- AGGATTCCTTGGCAACACGG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 43032339 - 43032361 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gpr45
- Entrez Gene ID
-
93690
- Gene Name
- G protein-coupled receptor 45
- Gene Aliases
-
9230112G11Rik, AW046772
- Chromosome Location
-
Chr.1: 42952872 - 43035451 on GRCm38
- UniGene ID
- Mm.444976
- Species
-
Mouse