gRNA Design Details
- Target DNA Sequence
- ACCCATACTTGACTTGGCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 56963095 - 56963117 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.47/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CETP
- Entrez Gene ID
-
1071
- Gene Name
- cholesteryl ester transfer protein
- Gene Aliases
-
BPIFF, HDLCQ10
- Chromosome Location
-
Chr.16: 56961923 - 56983844 on GRCh38
- UniGene ID
- Hs.89538
- Species
-
Human