gRNA Design Details

Target DNA Sequence
ATGCTACTGGCAACAGAACA
PAM Sequence
AGG
Target locus
Chr.8: 78798149 - 78798171 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.33/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IL7
Entrez Gene ID
3574
Gene Name
interleukin 7
Gene Aliases
IL-7
Chromosome Location
Chr.8: 78675870 - 78805523 on GRCh38
UniGene ID
Hs.591873
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000880.3 NP_000871.1 2
NM_001199886.1 NP_001186815.1 2
NM_001199887.1 NP_001186816.1 2
NM_001199888.1 NP_001186817.1 2
XM_011517522.2 XP_011515824.1 2
XM_011517523.2 XP_011515825.1 2
XM_017013397.1 XP_016868886.1 2