gRNA Design Details

Target DNA Sequence
GCGCTTCCCCCAGATAAATA
PAM Sequence
TGG
Target locus
Chr.6: 80007958 - 80007980 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 87.49/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TTK
Entrez Gene ID
7272
Gene Name
TTK protein kinase
Gene Aliases
CT96, ESK, MPH1, MPS1, MPS1L1, PYT
Chromosome Location
Chr.6: 80004147 - 80043577 on GRCh38
UniGene ID
Hs.169840
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001166691.1 NP_001160163.1 3
NM_003318.4 NP_003309.2 3
XM_011536099.2 XP_011534401.1 3
XM_011536100.2 XP_011534402.1 3
XM_017011242.1 XP_016866731.1 4