gRNA Design Details
- Target DNA Sequence
- AACACTTGTGGGGTGGTAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 41232812 - 41232834 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
81.58/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- KRTAP9-3
- Entrez Gene ID
-
83900
- Gene Name
- keratin associated protein 9-3
- Gene Aliases
-
KAP9.3, KRTAP9.3
- Chromosome Location
-
Chr.17: 41232463 - 41233454 on GRCh38
- UniGene ID
- Hs.307012
- Species
-
Human