gRNA Design Details
- Target DNA Sequence
- AATCTCCCTAACTGGCAGCG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 227815842 - 227815864 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS38
- Entrez Gene ID
-
339501
- Gene Name
- protease, serine 38
- Gene Aliases
-
MPN2
- Chromosome Location
-
Chr.1: 227815708 - 227846470 on GRCh38
- UniGene ID
- Hs.97604
- Species
-
Human