gRNA Design Details
- Target DNA Sequence
- GGAGTTTAGCCAGTCCAACT
- PAM Sequence
- CGG
- Target locus
-
Chr.13: 40666101 - 40666123 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FOXO1
- Entrez Gene ID
-
2308
- Gene Name
- forkhead box O1
- Gene Aliases
-
FKH1, FKHR, FOXO1A
- Chromosome Location
-
Chr.13: 40555664 - 40666597 on GRCh38
- UniGene ID
- Hs.370666
- Species
-
Human