gRNA Design Details
- Target DNA Sequence
- ATCGTGTGCACCATGCGAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 67487132 - 67487154 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AIP
- Entrez Gene ID
-
9049
- Gene Name
- aryl hydrocarbon receptor interacting protein
- Gene Aliases
-
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2
- Chromosome Location
-
Chr.11: 67483029 - 67491108 on GRCh38
- UniGene ID
- Hs.412433
- Species
-
Human