gRNA Design Details

Target DNA Sequence
ATCGTGTGCACCATGCGAGA
PAM Sequence
AGG
Target locus
Chr.11: 67487132 - 67487154 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 95.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AIP
Entrez Gene ID
9049
Gene Name
aryl hydrocarbon receptor interacting protein
Gene Aliases
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2
Chromosome Location
Chr.11: 67483029 - 67491108 on GRCh38
UniGene ID
Hs.412433
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001302959.1 NP_001289888.1 2
NM_001302960.1 NP_001289889.1 2
NM_003977.3 NP_003968.3 2