gRNA Design Details

Target DNA Sequence
GACTTCATCACAACCATTGG
PAM Sequence
AGG
Target locus
Chr.2: 237817032 - 237817054 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.61/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
RBM44
Entrez Gene ID
375316
Gene Name
RNA binding motif protein 44
Gene Aliases
-
Chromosome Location
Chr.2: 237798048 - 237842808 on GRCh38
UniGene ID
Hs.607265
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001080504.2 NP_001073973.2 3
XM_005246075.1 XP_005246132.1 3
XM_011511152.2 XP_011509454.1 3
XM_011511153.2 XP_011509455.1 3
XM_011511154.2 XP_011509456.1 3
XM_017004054.1 XP_016859543.1 3
XM_017004055.1 XP_016859544.1 3
XM_017004056.1 XP_016859545.1 3
XM_017004057.1 XP_016859546.1 3