Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001080504.2 | NP_001073973.2 | 2 |
XM_005246075.1 | XP_005246132.1 | 2 | |
XM_011511152.2 | XP_011509454.1 | 2 | |
XM_011511153.2 | XP_011509455.1 | 2 | |
XM_011511154.2 | XP_011509456.1 | 2 | |
XM_017004054.1 | XP_016859543.1 | 2 | |
XM_017004055.1 | XP_016859544.1 | 2 | |
XM_017004056.1 | XP_016859545.1 | 2 | |
XM_017004057.1 | XP_016859546.1 | 2 |