gRNA Design Details
- Target DNA Sequence
- AGAGTGGTTTCGGGCCTGTT
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 28681426 - 28681448 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BAMBI
- Entrez Gene ID
-
25805
- Gene Name
- BMP and activin membrane bound inhibitor
- Gene Aliases
-
NMA
- Chromosome Location
-
Chr.10: 28677495 - 28682939 on GRCh38
- UniGene ID
- Hs.533336
- Species
-
Human