gRNA Design Details

Target DNA Sequence
TTCCGGTCATCCCACTCTCC
PAM Sequence
AGG
Target locus
Chr.2: 218280051 - 218280073 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 87.39/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TMBIM1
Entrez Gene ID
64114
Gene Name
transmembrane BAX inhibitor motif containing 1
Gene Aliases
LFG3, MST100, MSTP100, PP1201, RECS1
Chromosome Location
Chr.2: 218274192 - 218292577 on GRCh38
UniGene ID
Hs.591605
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001321427.1 NP_001308356.1 4
NM_001321428.1 NP_001308357.1 3
NM_001321429.1 NP_001308358.1 4
NM_001321430.1 NP_001308359.1 4
NM_001321432.1 NP_001308361.1 3
NM_001321433.1 NP_001308362.1 4
NM_001321435.1 NP_001308364.1 4
NM_001321436.1 NP_001308365.1 4
NM_001321438.1 NP_001308367.1 3
NM_022152.5 NP_071435.2 3
NR_135643.1 2
XM_011511627.1 XP_011509929.1 3