gRNA Design Details
- Target DNA Sequence
- AGTGCCAATGTCGAGAACCA
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 68324747 - 68324769 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.15/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRMT7
- Entrez Gene ID
-
54496
- Gene Name
- protein arginine methyltransferase 7
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 68310974 - 68358832 on GRCh38
- UniGene ID
- Hs.640229
- Species
-
Human