Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199505.1 | NP_001186434.1 | 7 | 1090 | |
NM_001199506.1 | NP_001186435.1 | 7 | 1085 | ||
NM_001281507.1 | NP_001268436.1 | 7 | 1189 | ||
NM_080672.4 | NP_542403.1 | 7 | 1567 | ||
NM_183244.1 | NP_899067.1 | 7 | 1210 | ||
NM_183246.1 | NP_899069.1 | 6 | 1000 | ||
XM_011528525.2 | XP_011526827.1 | 7 | 1196 | ||
XM_011528526.2 | XP_011526828.1 | 6 | 1002 | ||
XM_017027626.1 | XP_016883115.1 | 7 | 1299 | ||
XM_017027627.1 | XP_016883116.1 | 7 | 1244 | ||
XM_017027628.1 | XP_016883117.1 | 6 | 892 | ||
XM_017027629.1 | XP_016883118.1 | 6 | 1035 | ||
XM_017027630.1 | XP_016883119.1 | 6 | 863 | ||
XM_017027631.1 | XP_016883120.1 | 7 | 1126 | ||
GenBank mRNA | AB098521.1 | - | 7 | 1210 | |
AB098522.1 | - | 6 | 1000 | ||
AJ311122.2 | - | 7 | 1566 | ||
AJ617581.1 | - | 7 | 1085 | ||
AK091632.1 | - | 7 | 1212 | ||
AK098788.1 | - | 7 | 1090 | ||
AK295459.1 | - | 7 | 1189 | ||
AK314493.1 | - | 7 | 1297 | ||
AK316047.1 | - | 7 | 1082 | ||
BC108303.1 | - | 7 | 1567 | 4179728 | |
BC117362.1 | - | 7 | 1455 | 40125913 | |
BC117364.1 | - | 7 | 1458 | 40125915 |