Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000157.3 | NP_000148.2 | 4 | 500 | |
NM_001005741.2 | NP_001005741.1 | 5 | 759 | ||
NM_001005742.2 | NP_001005742.1 | 5 | 740 | ||
NM_001171811.1 | NP_001165282.1 | 3 | 576 | ||
GenBank mRNA | AK291911.1 | - | 5 | 738 | |
AK300829.1 | - | 3 | 413 | ||
AK300876.1 | - | 5 | 740 | ||
AK302000.1 | - | 3 | 325 | ||
AK311242.1 | - | 5 | 348 | ||
AK312502.1 | - | 5 | 556 | ||
AL547558.3 | - | 6 | 671 | ||
AU122729.1 | - | 6 | 660 | ||
AU131071.1 | - | 5 | 568 | ||
AU140959.1 | - | 4 | 484 | ||
AU141794.1 | - | 5 | 575 | ||
BC003356.1 | - | 4 | 439 | 2899915 | |
BI458641.1 | - | 5 | 701 | ||
BX382279.2 | - | 6 | 658 | ||
D13286.1 | - | 4 | 439 | ||
DC297079.1 | - | 4 | 500 | ||
K02920.1 | - | 3 | 319 | ||
KJ690771.1 | - | 4 | 383 | ||
KJ690772.1 | - | 4 | 383 | ||
M16328.1 | - | 4 | 769 | ||
M19285.1 | - | 4 | 456 |