Gene Transcripts

Gene Symbol
TSPYL1
Entrez Gene ID
7259
Gene Name
TSPY like 1
Gene Aliases
TSPYL
Chromosome Location
Chr. 6: 116274859 - 116280117 on Build GRCh38
UniGene ID
Hs.458358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003309.3 NP_003300.1 1 1121
GenBank mRNA AL136629.1 - 1 935
BC048969.1 - 1 937 5297724
BC068554.1 - 1 934
CR533535.1 - 1 837

View Full Product Details