Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40559824-40560328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO1
Entrez Gene ID
2308
Gene Name
forkhead box O1
Gene Aliases
FKH1, FKHR, FOXO1A
UniGene
Hs.370666
Gene Chromosome Location
Chr.13: 40555664-40666597 on Build GRCh38
Transcripts
NM_002015.3 XM_011535008.2 XM_011535010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
356748 c.1360C>T p.Q454* Chr.13: 40560131-40560131 on Build GRCh38
6953250 c.1298delC p.P433fs*20 Chr.13: 40560193-40560193 on Build GRCh38
6976851 c.1343G>A p.S448N Chr.13: 40560148-40560148 on Build GRCh38
4622827 c.1325T>G p.L442R Chr.13: 40560166-40560166 on Build GRCh38
3793283 c.1241T>C p.L414S Chr.13: 40560250-40560250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760979256 Chr.13: 40559954-40559954 on Build GRCh38
rs927500436 Chr.13: 40559974-40559974 on Build GRCh38
rs771279465 Chr.13: 40560032-40560032 on Build GRCh38
rs751653854 Chr.13: 40560301-40560301 on Build GRCh38
rs747053864 Chr.13: 40560219-40560219 on Build GRCh38
rs767235452 Chr.13: 40560183-40560183 on Build GRCh38
rs754273701 Chr.13: 40559961-40559961 on Build GRCh38
rs570340177 Chr.13: 40559919-40559919 on Build GRCh38
rs70961707 Chr.13: 40559959-40559959 on Build GRCh38
rs753713529 Chr.13: 40560203-40560203 on Build GRCh38
rs148440850 Chr.13: 40559873-40559873 on Build GRCh38
rs191887079 Chr.13: 40559970-40559970 on Build GRCh38
rs200294842 Chr.13: 40560041-40560041 on Build GRCh38
rs762595333 Chr.13: 40560163-40560163 on Build GRCh38
rs1008631029 Chr.13: 40559872-40559872 on Build GRCh38
rs768903432 Chr.13: 40560020-40560020 on Build GRCh38
rs755802918 Chr.13: 40560196-40560196 on Build GRCh38
rs577834620 Chr.13: 40560182-40560182 on Build GRCh38
rs148177044 Chr.13: 40560286-40560286 on Build GRCh38
rs142437019 Chr.13: 40560156-40560156 on Build GRCh38
rs202007776 Chr.13: 40559976-40559976 on Build GRCh38
rs766344604 Chr.13: 40560200-40560200 on Build GRCh38
rs772361290 Chr.13: 40559950-40559950 on Build GRCh38
rs144016210 Chr.13: 40559881-40559881 on Build GRCh38
rs571854966 Chr.13: 40560133-40560133 on Build GRCh38
rs758562942 Chr.13: 40560208-40560208 on Build GRCh38
rs142210500 Chr.13: 40560024-40560024 on Build GRCh38
rs138145556 Chr.13: 40560134-40560134 on Build GRCh38
rs768256212 Chr.13: 40560264-40560264 on Build GRCh38
rs1005190964 Chr.13: 40560151-40560151 on Build GRCh38
rs778538205 Chr.13: 40559874-40559874 on Build GRCh38
rs1028000118 Chr.13: 40560130-40560130 on Build GRCh38
rs748268306 Chr.13: 40560091-40560091 on Build GRCh38
rs946277208 Chr.13: 40559996-40559996 on Build GRCh38
rs139264096 Chr.13: 40560257-40560257 on Build GRCh38
rs777178442 Chr.13: 40560036-40560036 on Build GRCh38
rs752387735 Chr.13: 40559878-40559878 on Build GRCh38
rs1038766455 Chr.13: 40560072-40560072 on Build GRCh38
rs144907712 Chr.13: 40560005-40560005 on Build GRCh38
rs146471778 Chr.13: 40560176-40560176 on Build GRCh38
rs751383976 Chr.13: 40559986-40559986 on Build GRCh38
rs199932286 Chr.13: 40559935-40559935 on Build GRCh38
rs573655708 Chr.13: 40560223-40560223 on Build GRCh38
rs991177035 Chr.13: 40559894-40559894 on Build GRCh38
rs757179771 Chr.13: 40560077-40560077 on Build GRCh38
rs146488369 Chr.13: 40559882-40559882 on Build GRCh38
rs752275319 Chr.13: 40560042-40560042 on Build GRCh38
rs34650827 Chr.13: 40559985-40559985 on Build GRCh38
rs757909015 Chr.13: 40560120-40560121 on Build GRCh38
rs750262452 Chr.13: 40560193-40560193 on Build GRCh38
rs758104682 Chr.13: 40559983-40559983 on Build GRCh38
rs374723626 Chr.13: 40560023-40560023 on Build GRCh38
rs143239174 Chr.13: 40560267-40560267 on Build GRCh38
rs753862295 Chr.13: 40560283-40560283 on Build GRCh38
rs762070851 Chr.13: 40559943-40559943 on Build GRCh38
rs765624020 Chr.13: 40560038-40560038 on Build GRCh38
rs750423870 Chr.13: 40560080-40560080 on Build GRCh38
rs369516414 Chr.13: 40560282-40560282 on Build GRCh38
rs144776359 Chr.13: 40560285-40560285 on Build GRCh38
rs181835164 Chr.13: 40560157-40560157 on Build GRCh38
rs779803394 Chr.13: 40560083-40560083 on Build GRCh38
rs774027114 Chr.13: 40560266-40560266 on Build GRCh38
rs757047359 Chr.13: 40559994-40559994 on Build GRCh38
rs557678402 Chr.13: 40560160-40560160 on Build GRCh38
rs777807232 Chr.13: 40560210-40560210 on Build GRCh38
rs372059765 Chr.13: 40559902-40559902 on Build GRCh38
rs151228712 Chr.13: 40560259-40560259 on Build GRCh38
rs766830396 Chr.13: 40559865-40559865 on Build GRCh38
rs763816192 Chr.13: 40560053-40560053 on Build GRCh38
rs776126179 Chr.13: 40560262-40560262 on Build GRCh38
rs151230081 Chr.13: 40560006-40560006 on Build GRCh38
rs759110513 Chr.13: 40559969-40559969 on Build GRCh38
rs762312415 Chr.13: 40560050-40560050 on Build GRCh38
rs145453868 Chr.13: 40559866-40559866 on Build GRCh38
rs778122381 Chr.13: 40560129-40560129 on Build GRCh38
rs867122605 Chr.13: 40559871-40559871 on Build GRCh38
rs202038357 Chr.13: 40559917-40559917 on Build GRCh38
rs369078513 Chr.13: 40560054-40560054 on Build GRCh38
rs772929365 Chr.13: 40560268-40560268 on Build GRCh38
rs774497137 Chr.13: 40559936-40559936 on Build GRCh38
rs767400829 Chr.13: 40560079-40560079 on Build GRCh38
rs140386357 Chr.13: 40560022-40560022 on Build GRCh38
rs771019295 Chr.13: 40559885-40559885 on Build GRCh38
rs201548760 Chr.13: 40560303-40560303 on Build GRCh38
rs201530379 Chr.13: 40560179-40560179 on Build GRCh38
rs548058902 Chr.13: 40559886-40559886 on Build GRCh38
rs371334379 Chr.13: 40560143-40560143 on Build GRCh38
rs994659376 Chr.13: 40560118-40560118 on Build GRCh38
rs950162499 Chr.13: 40559960-40559960 on Build GRCh38
rs112682206 Chr.13: 40559900-40559900 on Build GRCh38
rs777012971 Chr.13: 40560144-40560144 on Build GRCh38
rs768233462 Chr.13: 40559883-40559883 on Build GRCh38
rs760007713 Chr.13: 40560037-40560037 on Build GRCh38
rs757493915 Chr.13: 40560234-40560234 on Build GRCh38
rs760584890 Chr.13: 40560270-40560270 on Build GRCh38
rs555035704 Chr.13: 40560117-40560117 on Build GRCh38
rs890236166 Chr.13: 40559867-40559867 on Build GRCh38
rs749384551 Chr.13: 40559920-40559920 on Build GRCh38
rs12865456 Chr.13: 40560028-40560028 on Build GRCh38
rs781334894 Chr.13: 40560253-40560253 on Build GRCh38

View Full Product Details