Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92937227-92937719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs560889080 Chr.14: 92937476-92937476 on Build GRCh38
rs114404192 Chr.14: 92937421-92937421 on Build GRCh38
rs182660758 Chr.14: 92937404-92937404 on Build GRCh38
rs1040677699 Chr.14: 92937337-92937337 on Build GRCh38
rs2281514 Chr.14: 92937415-92937415 on Build GRCh38
rs116291263 Chr.14: 92937269-92937269 on Build GRCh38
rs976310085 Chr.14: 92937555-92937555 on Build GRCh38
rs995759736 Chr.14: 92937497-92937497 on Build GRCh38
rs753721071 Chr.14: 92937686-92937686 on Build GRCh38
rs1026700436 Chr.14: 92937510-92937510 on Build GRCh38
rs945054395 Chr.14: 92937680-92937680 on Build GRCh38
rs576069249 Chr.14: 92937313-92937313 on Build GRCh38
rs567481776 Chr.14: 92937662-92937662 on Build GRCh38
rs191287467 Chr.14: 92937388-92937388 on Build GRCh38
rs1044288365 Chr.14: 92937613-92937613 on Build GRCh38
rs945556516 Chr.14: 92937390-92937390 on Build GRCh38
rs1002173793 Chr.14: 92937425-92937425 on Build GRCh38
rs771604709 Chr.14: 92937308-92937308 on Build GRCh38
rs896523983 Chr.14: 92937484-92937484 on Build GRCh38
rs536438267 Chr.14: 92937667-92937667 on Build GRCh38
rs1027022832 Chr.14: 92937565-92937565 on Build GRCh38
rs556285518 Chr.14: 92937690-92937690 on Build GRCh38
rs959259054 Chr.14: 92937602-92937602 on Build GRCh38
rs551866479 Chr.14: 92937528-92937528 on Build GRCh38
rs527943694 Chr.14: 92937621-92937621 on Build GRCh38
rs961407285 Chr.14: 92937522-92937524 on Build GRCh38
rs952821385 Chr.14: 92937566-92937566 on Build GRCh38
rs919048801 Chr.14: 92937336-92937336 on Build GRCh38
rs984574821 Chr.14: 92937567-92937567 on Build GRCh38
rs992092422 Chr.14: 92937643-92937643 on Build GRCh38
rs1036879661 Chr.14: 92937698-92937698 on Build GRCh38
rs919688544 Chr.14: 92937263-92937263 on Build GRCh38
rs972842688 Chr.14: 92937325-92937325 on Build GRCh38
rs141349789 Chr.14: 92937342-92937342 on Build GRCh38
rs552715768 Chr.14: 92937624-92937624 on Build GRCh38
rs760385623 Chr.14: 92937461-92937461 on Build GRCh38
rs925511836 Chr.14: 92937622-92937622 on Build GRCh38
rs60511400 Chr.14: 92937490-92937490 on Build GRCh38
rs1015486961 Chr.14: 92937491-92937491 on Build GRCh38
rs1042487732 Chr.14: 92937689-92937689 on Build GRCh38
rs1013558012 Chr.14: 92937677-92937677 on Build GRCh38
rs1055164825 Chr.14: 92937665-92937665 on Build GRCh38
rs945532150 Chr.14: 92937608-92937608 on Build GRCh38
rs146992679 Chr.14: 92937495-92937495 on Build GRCh38
rs377094161 Chr.14: 92937304-92937304 on Build GRCh38
rs994607953 Chr.14: 92937524-92937524 on Build GRCh38
rs571941678 Chr.14: 92937553-92937553 on Build GRCh38
rs951872592 Chr.14: 92937338-92937338 on Build GRCh38
rs944933728 Chr.14: 92937309-92937309 on Build GRCh38
rs58190804 Chr.14: 92937287-92937287 on Build GRCh38
rs2281513 Chr.14: 92937521-92937521 on Build GRCh38
rs547974263 Chr.14: 92937653-92937653 on Build GRCh38
rs569494603 Chr.14: 92937631-92937631 on Build GRCh38
rs984626853 Chr.14: 92937339-92937339 on Build GRCh38
rs914173333 Chr.14: 92937557-92937557 on Build GRCh38
rs747571010 Chr.14: 92937405-92937405 on Build GRCh38
rs117559055 Chr.14: 92937511-92937511 on Build GRCh38
rs145601942 Chr.14: 92937576-92937577 on Build GRCh38
rs937958402 Chr.14: 92937374-92937374 on Build GRCh38

View Full Product Details