Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92940759-92941260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts
XM_017021262.1 XM_017021263.1 XM_017021264.1

SNP Detail

SNP ID Chromosome Location
rs904896924 Chr.14: 92940965-92940965 on Build GRCh38
rs895995220 Chr.14: 92940880-92940880 on Build GRCh38
rs967757208 Chr.14: 92941156-92941156 on Build GRCh38
rs567194140 Chr.14: 92940886-92940886 on Build GRCh38
rs994906956 Chr.14: 92941212-92941212 on Build GRCh38
rs1015435190 Chr.14: 92940899-92940899 on Build GRCh38
rs1014968814 Chr.14: 92941052-92941052 on Build GRCh38
rs540239378 Chr.14: 92940783-92940783 on Build GRCh38
rs994958413 Chr.14: 92941068-92941068 on Build GRCh38
rs1043255239 Chr.14: 92940862-92940862 on Build GRCh38
rs776501911 Chr.14: 92940821-92940821 on Build GRCh38
rs889093666 Chr.14: 92941227-92941227 on Build GRCh38
rs911712232 Chr.14: 92940942-92940942 on Build GRCh38
rs573599711 Chr.14: 92941102-92941102 on Build GRCh38
rs980832875 Chr.14: 92941182-92941182 on Build GRCh38
rs746444980 Chr.14: 92940795-92940795 on Build GRCh38
rs558641660 Chr.14: 92941180-92941180 on Build GRCh38
rs145051520 Chr.14: 92940793-92940794 on Build GRCh38
rs968779201 Chr.14: 92941067-92941067 on Build GRCh38
rs551493348 Chr.14: 92940854-92940854 on Build GRCh38
rs1044213194 Chr.14: 92940853-92940853 on Build GRCh38
rs946399759 Chr.14: 92940857-92940857 on Build GRCh38
rs140475831 Chr.14: 92941072-92941072 on Build GRCh38
rs933561477 Chr.14: 92940788-92940788 on Build GRCh38
rs763123467 Chr.14: 92940951-92940951 on Build GRCh38
rs181295496 Chr.14: 92941104-92941104 on Build GRCh38
rs753274672 Chr.14: 92941090-92941090 on Build GRCh38
rs532360145 Chr.14: 92940816-92940816 on Build GRCh38
rs578020513 Chr.14: 92940998-92940998 on Build GRCh38
rs77138997 Chr.14: 92940902-92940902 on Build GRCh38
rs917491595 Chr.14: 92941176-92941176 on Build GRCh38
rs150435643 Chr.14: 92941109-92941109 on Build GRCh38
rs1050520143 Chr.14: 92941215-92941215 on Build GRCh38
rs77136775 Chr.14: 92941161-92941161 on Build GRCh38
rs11446 Chr.14: 92940903-92940903 on Build GRCh38
rs895954808 Chr.14: 92941035-92941035 on Build GRCh38
rs933748501 Chr.14: 92940907-92940907 on Build GRCh38
rs775964822 Chr.14: 92940948-92940948 on Build GRCh38
rs953269884 Chr.14: 92940910-92940910 on Build GRCh38
rs1044709091 Chr.14: 92940964-92940964 on Build GRCh38
rs78863877 Chr.14: 92941160-92941160 on Build GRCh38
rs561685992 Chr.14: 92940875-92940875 on Build GRCh38
rs767689662 Chr.14: 92940963-92940963 on Build GRCh38
rs11160096 Chr.14: 92941136-92941136 on Build GRCh38
rs987825740 Chr.14: 92940941-92940941 on Build GRCh38
rs1005467645 Chr.14: 92941051-92941051 on Build GRCh38
rs915115915 Chr.14: 92941142-92941142 on Build GRCh38
rs35502373 Chr.14: 92940789-92940789 on Build GRCh38
rs925585421 Chr.14: 92940823-92940823 on Build GRCh38
rs940954903 Chr.14: 92940924-92940924 on Build GRCh38
rs142097656 Chr.14: 92940850-92940851 on Build GRCh38
rs147625288 Chr.14: 92941022-92941022 on Build GRCh38
rs13316 Chr.14: 92940956-92940956 on Build GRCh38
rs45503595 Chr.14: 92941238-92941238 on Build GRCh38
rs980513188 Chr.14: 92941158-92941158 on Build GRCh38
rs3742711 Chr.14: 92941190-92941190 on Build GRCh38
rs1037627085 Chr.14: 92940929-92940929 on Build GRCh38
rs538211199 Chr.14: 92941166-92941166 on Build GRCh38
rs899242946 Chr.14: 92941053-92941053 on Build GRCh38
rs12588550 Chr.14: 92941141-92941141 on Build GRCh38
rs186127464 Chr.14: 92940865-92940865 on Build GRCh38
rs1003115467 Chr.14: 92940996-92940996 on Build GRCh38
rs891680681 Chr.14: 92941005-92941005 on Build GRCh38
rs548786417 Chr.14: 92940790-92940790 on Build GRCh38
rs535798296 Chr.14: 92940822-92940822 on Build GRCh38
rs1052638483 Chr.14: 92940830-92940830 on Build GRCh38
rs908127449 Chr.14: 92940923-92940923 on Build GRCh38
rs533819460 Chr.14: 92940871-92940871 on Build GRCh38
rs560048364 Chr.14: 92940784-92940784 on Build GRCh38
rs113099282 Chr.14: 92940840-92940840 on Build GRCh38
rs114496241 Chr.14: 92941210-92941210 on Build GRCh38
rs773203830 Chr.14: 92940864-92940864 on Build GRCh38
rs562293795 Chr.14: 92940794-92940794 on Build GRCh38
rs1021273614 Chr.14: 92941066-92941066 on Build GRCh38
rs538054519 Chr.14: 92940961-92940961 on Build GRCh38
rs185601690 Chr.14: 92941189-92941189 on Build GRCh38
rs753692871 Chr.14: 92940973-92940973 on Build GRCh38
rs770034432 Chr.14: 92940797-92940797 on Build GRCh38
rs373810322 Chr.14: 92940909-92940909 on Build GRCh38
rs1042977892 Chr.14: 92941220-92941220 on Build GRCh38
rs386780076 Chr.14: 92940902-92940903 on Build GRCh38
rs996237464 Chr.14: 92941237-92941237 on Build GRCh38
rs1057349753 Chr.14: 92941168-92941168 on Build GRCh38
rs940694317 Chr.14: 92940791-92940791 on Build GRCh38
rs1056944483 Chr.14: 92941203-92941203 on Build GRCh38
rs189306468 Chr.14: 92940962-92940962 on Build GRCh38

View Full Product Details