Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49423100-49423611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
UniGene
Hs.567268
Gene Chromosome Location
Chr.15: 49423178-49487326 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

SNP Detail

SNP ID Chromosome Location
rs531048110 Chr.15: 49423379-49423379 on Build GRCh38
rs920060589 Chr.15: 49423167-49423167 on Build GRCh38
rs779480450 Chr.15: 49423123-49423124 on Build GRCh38
rs995552999 Chr.15: 49423490-49423490 on Build GRCh38
rs559608834 Chr.15: 49423357-49423357 on Build GRCh38
rs930000113 Chr.15: 49423312-49423312 on Build GRCh38
rs1015519665 Chr.15: 49423145-49423145 on Build GRCh38
rs541994338 Chr.15: 49423367-49423367 on Build GRCh38
rs576716418 Chr.15: 49423188-49423188 on Build GRCh38
rs900216765 Chr.15: 49423307-49423307 on Build GRCh38
rs1043191246 Chr.15: 49423281-49423281 on Build GRCh38
rs754546768 Chr.15: 49423275-49423276 on Build GRCh38
rs73398286 Chr.15: 49423192-49423192 on Build GRCh38
rs1038453602 Chr.15: 49423297-49423297 on Build GRCh38
rs980608893 Chr.15: 49423574-49423574 on Build GRCh38
rs967022880 Chr.15: 49423539-49423539 on Build GRCh38
rs933140532 Chr.15: 49423195-49423195 on Build GRCh38
rs985949414 Chr.15: 49423578-49423578 on Build GRCh38
rs564510516 Chr.15: 49423515-49423515 on Build GRCh38
rs894195630 Chr.15: 49423419-49423419 on Build GRCh38
rs986388753 Chr.15: 49423199-49423199 on Build GRCh38
rs961387443 Chr.15: 49423160-49423160 on Build GRCh38
rs928472910 Chr.15: 49423218-49423218 on Build GRCh38
rs563473641 Chr.15: 49423377-49423377 on Build GRCh38
rs544623725 Chr.15: 49423343-49423343 on Build GRCh38
rs193188060 Chr.15: 49423562-49423562 on Build GRCh38
rs1053054999 Chr.15: 49423230-49423230 on Build GRCh38
rs967075304 Chr.15: 49423164-49423164 on Build GRCh38
rs947112509 Chr.15: 49423265-49423265 on Build GRCh38
rs1021308237 Chr.15: 49423146-49423146 on Build GRCh38
rs974542687 Chr.15: 49423161-49423161 on Build GRCh38
rs1015822262 Chr.15: 49423410-49423410 on Build GRCh38
rs762511219 Chr.15: 49423452-49423455 on Build GRCh38
rs553628239 Chr.15: 49423385-49423385 on Build GRCh38
rs562566267 Chr.15: 49423554-49423554 on Build GRCh38
rs574735445 Chr.15: 49423359-49423359 on Build GRCh38
rs143077465 Chr.15: 49423558-49423559 on Build GRCh38
rs938553865 Chr.15: 49423224-49423224 on Build GRCh38
rs979924372 Chr.15: 49423196-49423196 on Build GRCh38
rs897157724 Chr.15: 49423435-49423435 on Build GRCh38
rs912887199 Chr.15: 49423247-49423247 on Build GRCh38

View Full Product Details