Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49424309-49424812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
UniGene
Hs.567268
Gene Chromosome Location
Chr.15: 49423178-49487326 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
962478 c.131T>C p.V44A Chr.15: 49424428-49424428 on Build GRCh38
962479 c.161G>T p.R54I Chr.15: 49424458-49424458 on Build GRCh38
5722313 c.70C>T p.L24L Chr.15: 49424367-49424367 on Build GRCh38
4055403 c.238G>A p.D80N Chr.15: 49424535-49424535 on Build GRCh38
433998 c.115C>T p.Q39* Chr.15: 49424412-49424412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746255850 Chr.15: 49424400-49424400 on Build GRCh38
rs777358667 Chr.15: 49424636-49424636 on Build GRCh38
rs765581354 Chr.15: 49424591-49424592 on Build GRCh38
rs746522643 Chr.15: 49424432-49424432 on Build GRCh38
rs764963094 Chr.15: 49424334-49424334 on Build GRCh38
rs768087610 Chr.15: 49424438-49424438 on Build GRCh38
rs764278568 Chr.15: 49424374-49424374 on Build GRCh38
rs758913734 Chr.15: 49424404-49424404 on Build GRCh38
rs1042215908 Chr.15: 49424527-49424527 on Build GRCh38
rs779375910 Chr.15: 49424398-49424398 on Build GRCh38
rs16962440 Chr.15: 49424648-49424648 on Build GRCh38
rs753997634 Chr.15: 49424552-49424552 on Build GRCh38
rs747707846 Chr.15: 49424628-49424628 on Build GRCh38
rs754058875 Chr.15: 49424376-49424376 on Build GRCh38
rs951838150 Chr.15: 49424737-49424737 on Build GRCh38
rs764225350 Chr.15: 49424550-49424550 on Build GRCh38
rs750784831 Chr.15: 49424596-49424596 on Build GRCh38
rs377247133 Chr.15: 49424612-49424612 on Build GRCh38
rs769477922 Chr.15: 49424486-49424486 on Build GRCh38
rs71467679 Chr.15: 49424619-49424619 on Build GRCh38
rs370236693 Chr.15: 49424502-49424502 on Build GRCh38
rs559719143 Chr.15: 49424671-49424671 on Build GRCh38
rs1057133325 Chr.15: 49424414-49424414 on Build GRCh38
rs376909937 Chr.15: 49424418-49424418 on Build GRCh38
rs895834784 Chr.15: 49424422-49424422 on Build GRCh38
rs762071927 Chr.15: 49424584-49424584 on Build GRCh38
rs766821099 Chr.15: 49424631-49424632 on Build GRCh38
rs529145599 Chr.15: 49424504-49424504 on Build GRCh38
rs762092181 Chr.15: 49424581-49424581 on Build GRCh38
rs971786084 Chr.15: 49424725-49424725 on Build GRCh38
rs1010311644 Chr.15: 49424468-49424468 on Build GRCh38
rs961712096 Chr.15: 49424717-49424717 on Build GRCh38
rs772911643 Chr.15: 49424341-49424341 on Build GRCh38
rs763200068 Chr.15: 49424625-49424626 on Build GRCh38
rs183624878 Chr.15: 49424519-49424519 on Build GRCh38
rs766278075 Chr.15: 49424365-49424365 on Build GRCh38
rs780650709 Chr.15: 49424618-49424618 on Build GRCh38
rs752811080 Chr.15: 49424366-49424366 on Build GRCh38
rs1018535842 Chr.15: 49424676-49424676 on Build GRCh38
rs757572324 Chr.15: 49424379-49424379 on Build GRCh38
rs755511380 Chr.15: 49424614-49424614 on Build GRCh38
rs780470400 Chr.15: 49424606-49424606 on Build GRCh38
rs890102153 Chr.15: 49424340-49424340 on Build GRCh38
rs377679450 Chr.15: 49424342-49424342 on Build GRCh38
rs984658938 Chr.15: 49424764-49424764 on Build GRCh38
rs528536819 Chr.15: 49424604-49424604 on Build GRCh38
rs747759102 Chr.15: 49424483-49424483 on Build GRCh38
rs756216899 Chr.15: 49424367-49424367 on Build GRCh38
rs1000468210 Chr.15: 49424632-49424632 on Build GRCh38
rs750764359 Chr.15: 49424623-49424623 on Build GRCh38
rs112975672 Chr.15: 49424735-49424735 on Build GRCh38
rs745689304 Chr.15: 49424489-49424489 on Build GRCh38
rs34531231 Chr.15: 49424473-49424473 on Build GRCh38
rs776131808 Chr.15: 49424472-49424472 on Build GRCh38
rs376912968 Chr.15: 49424739-49424739 on Build GRCh38
rs774091678 Chr.15: 49424537-49424537 on Build GRCh38
rs766338451 Chr.15: 49424511-49424511 on Build GRCh38
rs373498840 Chr.15: 49424381-49424381 on Build GRCh38
rs928765064 Chr.15: 49424680-49424680 on Build GRCh38
rs143883386 Chr.15: 49424542-49424542 on Build GRCh38
rs17850705 Chr.15: 49424482-49424482 on Build GRCh38
rs375963598 Chr.15: 49424586-49424586 on Build GRCh38
rs780482125 Chr.15: 49424420-49424420 on Build GRCh38
rs1017443702 Chr.15: 49424382-49424382 on Build GRCh38

View Full Product Details