Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49485091-49485595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
UniGene
Hs.567268
Gene Chromosome Location
Chr.15: 49423178-49487326 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

SNP Detail

SNP ID Chromosome Location
rs892003999 Chr.15: 49485411-49485411 on Build GRCh38
rs371359059 Chr.15: 49485330-49485330 on Build GRCh38
rs980499556 Chr.15: 49485558-49485558 on Build GRCh38
rs12787 Chr.15: 49485339-49485339 on Build GRCh38
rs530204047 Chr.15: 49485556-49485556 on Build GRCh38
rs1027351117 Chr.15: 49485479-49485479 on Build GRCh38
rs112843832 Chr.15: 49485207-49485207 on Build GRCh38
rs1057657 Chr.15: 49485474-49485474 on Build GRCh38
rs960536767 Chr.15: 49485522-49485522 on Build GRCh38
rs1004842286 Chr.15: 49485493-49485493 on Build GRCh38
rs182077617 Chr.15: 49485400-49485400 on Build GRCh38
rs1040536229 Chr.15: 49485464-49485464 on Build GRCh38
rs1057656 Chr.15: 49485465-49485465 on Build GRCh38
rs12021 Chr.15: 49485417-49485417 on Build GRCh38
rs944420418 Chr.15: 49485443-49485443 on Build GRCh38
rs996225917 Chr.15: 49485474-49485475 on Build GRCh38
rs1064338 Chr.15: 49485568-49485568 on Build GRCh38
rs887685925 Chr.15: 49485483-49485483 on Build GRCh38
rs201990168 Chr.15: 49485234-49485235 on Build GRCh38
rs961449664 Chr.15: 49485224-49485224 on Build GRCh38
rs79465035 Chr.15: 49485474-49485474 on Build GRCh38
rs1057655 Chr.15: 49485445-49485445 on Build GRCh38
rs544082512 Chr.15: 49485288-49485288 on Build GRCh38
rs60927272 Chr.15: 49485236-49485237 on Build GRCh38
rs535504301 Chr.15: 49485474-49485475 on Build GRCh38
rs183166589 Chr.15: 49485552-49485552 on Build GRCh38
rs936288646 Chr.15: 49485378-49485378 on Build GRCh38
rs1014798713 Chr.15: 49485178-49485178 on Build GRCh38
rs983271082 Chr.15: 49485177-49485177 on Build GRCh38
rs948740950 Chr.15: 49485354-49485354 on Build GRCh38
rs3208783 Chr.15: 49485126-49485126 on Build GRCh38
rs185098313 Chr.15: 49485434-49485434 on Build GRCh38
rs980358392 Chr.15: 49485371-49485371 on Build GRCh38
rs190085424 Chr.15: 49485512-49485512 on Build GRCh38
rs544778626 Chr.15: 49485551-49485551 on Build GRCh38
rs926181021 Chr.15: 49485377-49485377 on Build GRCh38
rs60771113 Chr.15: 49485271-49485271 on Build GRCh38
rs35073573 Chr.15: 49485295-49485296 on Build GRCh38

View Full Product Details