Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27344779-27345284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_011545825.1 XM_011545826.2 XM_011545827.2 XM_011545828.2 XM_011545829.1 XM_011545830.2 XM_011545831.1 XM_011545832.1 XM_011545833.2 XM_011545834.2 XM_017023211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5619987 c.232A>T p.N78Y Chr.16: 27344891-27344891 on Build GRCh38
7127136 c.337G>T p.G113C Chr.16: 27344996-27344996 on Build GRCh38
1210599 c.218C>T p.T73M Chr.16: 27344877-27344877 on Build GRCh38
6397015 c.317G>A p.G106E Chr.16: 27344976-27344976 on Build GRCh38
6294584 c.253G>T p.V85L Chr.16: 27344912-27344912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56411840 Chr.16: 27345140-27345140 on Build GRCh38
rs554970103 Chr.16: 27344857-27344857 on Build GRCh38
rs757370872 Chr.16: 27345163-27345163 on Build GRCh38
rs761781246 Chr.16: 27344952-27344952 on Build GRCh38
rs766157629 Chr.16: 27345061-27345061 on Build GRCh38
rs543167942 Chr.16: 27345000-27345000 on Build GRCh38
rs893287811 Chr.16: 27344816-27344816 on Build GRCh38
rs888973268 Chr.16: 27345047-27345047 on Build GRCh38
rs960324104 Chr.16: 27345081-27345081 on Build GRCh38
rs375159221 Chr.16: 27344921-27344921 on Build GRCh38
rs1805010 Chr.16: 27344882-27344882 on Build GRCh38
rs750189532 Chr.16: 27344960-27344960 on Build GRCh38
rs765758630 Chr.16: 27345203-27345203 on Build GRCh38
rs145184963 Chr.16: 27344896-27344896 on Build GRCh38
rs753929235 Chr.16: 27345010-27345010 on Build GRCh38
rs752792692 Chr.16: 27344909-27344909 on Build GRCh38
rs750194290 Chr.16: 27344904-27344904 on Build GRCh38
rs962166185 Chr.16: 27345210-27345210 on Build GRCh38
rs113886028 Chr.16: 27344862-27344862 on Build GRCh38
rs144651842 Chr.16: 27344903-27344903 on Build GRCh38
rs141983833 Chr.16: 27344912-27344912 on Build GRCh38
rs781298536 Chr.16: 27344906-27344906 on Build GRCh38
rs759162058 Chr.16: 27344880-27344880 on Build GRCh38
rs1034710194 Chr.16: 27345053-27345053 on Build GRCh38
rs775052929 Chr.16: 27344942-27344942 on Build GRCh38
rs984623419 Chr.16: 27345202-27345202 on Build GRCh38
rs991472366 Chr.16: 27344999-27344999 on Build GRCh38
rs775017923 Chr.16: 27344887-27344887 on Build GRCh38
rs1035376706 Chr.16: 27344879-27344879 on Build GRCh38
rs570293541 Chr.16: 27345133-27345133 on Build GRCh38
rs770243204 Chr.16: 27344826-27344826 on Build GRCh38
rs565620025 Chr.16: 27345070-27345070 on Build GRCh38
rs375589999 Chr.16: 27345112-27345112 on Build GRCh38
rs1022034958 Chr.16: 27344856-27344856 on Build GRCh38
rs569547141 Chr.16: 27344878-27344878 on Build GRCh38
rs765284416 Chr.16: 27344959-27344959 on Build GRCh38
rs758746097 Chr.16: 27345019-27345019 on Build GRCh38
rs996376416 Chr.16: 27344803-27344803 on Build GRCh38
rs764325424 Chr.16: 27345150-27345150 on Build GRCh38
rs771597376 Chr.16: 27345042-27345042 on Build GRCh38
rs367548039 Chr.16: 27344902-27344902 on Build GRCh38
rs759026211 Chr.16: 27345225-27345225 on Build GRCh38
rs746593366 Chr.16: 27345032-27345032 on Build GRCh38
rs748195646 Chr.16: 27344819-27344819 on Build GRCh38
rs574446159 Chr.16: 27344998-27344998 on Build GRCh38
rs369179205 Chr.16: 27345138-27345138 on Build GRCh38
rs762694127 Chr.16: 27344993-27344993 on Build GRCh38
rs999545268 Chr.16: 27344817-27344817 on Build GRCh38
rs530143808 Chr.16: 27344804-27344804 on Build GRCh38
rs17548711 Chr.16: 27345093-27345093 on Build GRCh38
rs1050702653 Chr.16: 27345251-27345251 on Build GRCh38
rs779751751 Chr.16: 27345031-27345031 on Build GRCh38
rs1008636293 Chr.16: 27345055-27345055 on Build GRCh38
rs897471715 Chr.16: 27345089-27345089 on Build GRCh38
rs776228073 Chr.16: 27345039-27345039 on Build GRCh38
rs764099093 Chr.16: 27344897-27344897 on Build GRCh38
rs576739360 Chr.16: 27345012-27345012 on Build GRCh38
rs755176173 Chr.16: 27344905-27344905 on Build GRCh38
rs77491333 Chr.16: 27345043-27345043 on Build GRCh38
rs750955526 Chr.16: 27345213-27345213 on Build GRCh38
rs569508584 Chr.16: 27345215-27345215 on Build GRCh38
rs778294946 Chr.16: 27344834-27344834 on Build GRCh38
rs779320253 Chr.16: 27345013-27345013 on Build GRCh38
rs1012128011 Chr.16: 27345082-27345082 on Build GRCh38
rs746241652 Chr.16: 27344935-27344935 on Build GRCh38
rs931959761 Chr.16: 27345229-27345229 on Build GRCh38
rs1050629895 Chr.16: 27344986-27344986 on Build GRCh38
rs902582750 Chr.16: 27344809-27344809 on Build GRCh38
rs778243651 Chr.16: 27344916-27344916 on Build GRCh38
rs112497527 Chr.16: 27344950-27344950 on Build GRCh38
rs368631624 Chr.16: 27344926-27344926 on Build GRCh38
rs2072130 Chr.16: 27345077-27345077 on Build GRCh38
rs369875568 Chr.16: 27345044-27345044 on Build GRCh38
rs2074571 Chr.16: 27345091-27345091 on Build GRCh38
rs1010665730 Chr.16: 27344833-27344833 on Build GRCh38
rs141903283 Chr.16: 27345014-27345014 on Build GRCh38
rs753878304 Chr.16: 27345161-27345161 on Build GRCh38
rs773075015 Chr.16: 27345048-27345048 on Build GRCh38
rs1011518566 Chr.16: 27345092-27345092 on Build GRCh38
rs760218604 Chr.16: 27344946-27344946 on Build GRCh38
rs745518329 Chr.16: 27344859-27344859 on Build GRCh38
rs1011903425 Chr.16: 27344890-27344890 on Build GRCh38
rs199703000 Chr.16: 27345002-27345002 on Build GRCh38
rs2074572 Chr.16: 27345038-27345038 on Build GRCh38
rs1049657568 Chr.16: 27345127-27345127 on Build GRCh38
rs372171065 Chr.16: 27344917-27344917 on Build GRCh38
rs771717750 Chr.16: 27344936-27344936 on Build GRCh38
rs907951146 Chr.16: 27344907-27344907 on Build GRCh38
rs55832501 Chr.16: 27345049-27345049 on Build GRCh38

View Full Product Details