Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87504312-87504808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MINPP1
Entrez Gene ID
9562
Gene Name
multiple inositol-polyphosphate phosphatase 1
Gene Aliases
HIPER1, MINPP2, MIPP
UniGene
Hs.121260
Gene Chromosome Location
Chr.10: 87502792-87553461 on Build GRCh38
Transcripts
NM_001178117.1 NM_004897.4 XM_006718078.2 XM_017016965.1

SNP Detail

SNP ID Chromosome Location
rs555934932 Chr.10: 87504788-87504788 on Build GRCh38
rs1010689735 Chr.10: 87504630-87504630 on Build GRCh38
rs1014121777 Chr.10: 87504567-87504567 on Build GRCh38
rs937875333 Chr.10: 87504777-87504777 on Build GRCh38
rs921351754 Chr.10: 87504513-87504513 on Build GRCh38
rs74150471 Chr.10: 87504673-87504673 on Build GRCh38
rs868139220 Chr.10: 87504421-87504421 on Build GRCh38
rs913332252 Chr.10: 87504535-87504535 on Build GRCh38
rs952992942 Chr.10: 87504731-87504731 on Build GRCh38
rs1042617071 Chr.10: 87504707-87504707 on Build GRCh38
rs373715768 Chr.10: 87504768-87504768 on Build GRCh38
rs567247277 Chr.10: 87504711-87504711 on Build GRCh38
rs965561580 Chr.10: 87504713-87504713 on Build GRCh38
rs548670112 Chr.10: 87504695-87504695 on Build GRCh38
rs1007202138 Chr.10: 87504676-87504676 on Build GRCh38
rs965532858 Chr.10: 87504445-87504445 on Build GRCh38
rs2233163 Chr.10: 87504564-87504564 on Build GRCh38
rs919423746 Chr.10: 87504752-87504752 on Build GRCh38
rs755179663 Chr.10: 87504619-87504619 on Build GRCh38
rs562419653 Chr.10: 87504387-87504387 on Build GRCh38
rs1021119877 Chr.10: 87504647-87504647 on Build GRCh38
rs2233164 Chr.10: 87504719-87504719 on Build GRCh38
rs972240777 Chr.10: 87504751-87504751 on Build GRCh38
rs113131475 Chr.10: 87504528-87504528 on Build GRCh38
rs982083228 Chr.10: 87504773-87504773 on Build GRCh38
rs1049100423 Chr.10: 87504469-87504469 on Build GRCh38
rs1032727278 Chr.10: 87504650-87504650 on Build GRCh38
rs972186938 Chr.10: 87504349-87504349 on Build GRCh38
rs867178580 Chr.10: 87504479-87504479 on Build GRCh38
rs930721871 Chr.10: 87504419-87504419 on Build GRCh38
rs148523706 Chr.10: 87504566-87504566 on Build GRCh38
rs112694774 Chr.10: 87504532-87504532 on Build GRCh38
rs2233165 Chr.10: 87504774-87504774 on Build GRCh38
rs937995309 Chr.10: 87504520-87504520 on Build GRCh38
rs977681549 Chr.10: 87504403-87504404 on Build GRCh38
rs940014644 Chr.10: 87504771-87504771 on Build GRCh38
rs906418322 Chr.10: 87504607-87504607 on Build GRCh38
rs191762158 Chr.10: 87504359-87504359 on Build GRCh38
rs1041563525 Chr.10: 87504575-87504575 on Build GRCh38
rs927869806 Chr.10: 87504710-87504710 on Build GRCh38
rs562777365 Chr.10: 87504595-87504595 on Build GRCh38
rs940000086 Chr.10: 87504587-87504587 on Build GRCh38
rs1019812129 Chr.10: 87504431-87504431 on Build GRCh38
rs1046373141 Chr.10: 87504588-87504588 on Build GRCh38
rs902972695 Chr.10: 87504610-87504610 on Build GRCh38
rs946522355 Chr.10: 87504786-87504786 on Build GRCh38
rs766846208 Chr.10: 87504547-87504547 on Build GRCh38
rs370016679 Chr.10: 87504679-87504679 on Build GRCh38
rs915117981 Chr.10: 87504783-87504783 on Build GRCh38
rs756423993 Chr.10: 87504780-87504780 on Build GRCh38
rs1028526757 Chr.10: 87504730-87504730 on Build GRCh38
rs551503476 Chr.10: 87504623-87504623 on Build GRCh38
rs550087887 Chr.10: 87504725-87504725 on Build GRCh38
rs2233162 Chr.10: 87504496-87504496 on Build GRCh38
rs984606323 Chr.10: 87504548-87504548 on Build GRCh38
rs893554144 Chr.10: 87504628-87504628 on Build GRCh38
rs183634732 Chr.10: 87504617-87504617 on Build GRCh38

View Full Product Details