Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87508235-87508785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MINPP1
Entrez Gene ID
9562
Gene Name
multiple inositol-polyphosphate phosphatase 1
Gene Aliases
HIPER1, MINPP2, MIPP
UniGene
Hs.121260
Gene Chromosome Location
Chr.10: 87502792-87553461 on Build GRCh38
Transcripts
NM_001178117.1 NM_001178118.1 NM_004897.4 XM_006718078.2 XM_011540379.2 XM_017016965.1 XM_017016966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415424 c.697G>C p.E233Q Chr.10: 87508395-87508395 on Build GRCh38
7327931 c.75G>A p.M25I Chr.10: 87508376-87508376 on Build GRCh38
7327930 c.678G>A p.M226I Chr.10: 87508376-87508376 on Build GRCh38
4505390 c.691C>T p.H231Y Chr.10: 87508389-87508389 on Build GRCh38
1317419 c.744C>T p.H248H Chr.10: 87508442-87508442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537372139 Chr.10: 87508599-87508603 on Build GRCh38
rs968461177 Chr.10: 87508703-87508703 on Build GRCh38
rs202158684 Chr.10: 87508343-87508343 on Build GRCh38
rs758509432 Chr.10: 87508388-87508388 on Build GRCh38
rs910900695 Chr.10: 87508755-87508755 on Build GRCh38
rs1026420496 Chr.10: 87508443-87508443 on Build GRCh38
rs980450324 Chr.10: 87508646-87508646 on Build GRCh38
rs1051242120 Chr.10: 87508440-87508440 on Build GRCh38
rs748063255 Chr.10: 87508438-87508438 on Build GRCh38
rs780326945 Chr.10: 87508409-87508409 on Build GRCh38
rs762467837 Chr.10: 87508328-87508328 on Build GRCh38
rs541767127 Chr.10: 87508536-87508536 on Build GRCh38
rs770472841 Chr.10: 87508566-87508566 on Build GRCh38
rs759179191 Chr.10: 87508472-87508472 on Build GRCh38
rs905651989 Chr.10: 87508625-87508625 on Build GRCh38
rs777781974 Chr.10: 87508540-87508540 on Build GRCh38
rs774041685 Chr.10: 87508470-87508470 on Build GRCh38
rs1020925817 Chr.10: 87508699-87508699 on Build GRCh38
rs565257221 Chr.10: 87508704-87508704 on Build GRCh38
rs146087494 Chr.10: 87508657-87508657 on Build GRCh38
rs201618662 Chr.10: 87508459-87508459 on Build GRCh38
rs912990025 Chr.10: 87508674-87508674 on Build GRCh38
rs769420415 Chr.10: 87508321-87508321 on Build GRCh38
rs758694689 Chr.10: 87508296-87508296 on Build GRCh38
rs575102965 Chr.10: 87508655-87508655 on Build GRCh38
rs534346216 Chr.10: 87508308-87508308 on Build GRCh38
rs940293465 Chr.10: 87508456-87508456 on Build GRCh38
rs778328377 Chr.10: 87508575-87508575 on Build GRCh38
rs190555886 Chr.10: 87508544-87508544 on Build GRCh38
rs762348571 Chr.10: 87508357-87508357 on Build GRCh38
rs768977489 Chr.10: 87508484-87508484 on Build GRCh38
rs746047331 Chr.10: 87508286-87508286 on Build GRCh38
rs986562840 Chr.10: 87508749-87508749 on Build GRCh38
rs777031012 Chr.10: 87508492-87508492 on Build GRCh38
rs772885921 Chr.10: 87508324-87508324 on Build GRCh38
rs368274378 Chr.10: 87508316-87508316 on Build GRCh38
rs751799548 Chr.10: 87508524-87508524 on Build GRCh38
rs147897043 Chr.10: 87508506-87508506 on Build GRCh38
rs779664532 Chr.10: 87508299-87508299 on Build GRCh38
rs898171357 Chr.10: 87508519-87508519 on Build GRCh38
rs747702345 Chr.10: 87508450-87508450 on Build GRCh38
rs781108940 Chr.10: 87508531-87508531 on Build GRCh38
rs765763863 Chr.10: 87508497-87508497 on Build GRCh38
rs933775004 Chr.10: 87508695-87508695 on Build GRCh38
rs371951706 Chr.10: 87508333-87508333 on Build GRCh38
rs748152849 Chr.10: 87508319-87508319 on Build GRCh38
rs913916487 Chr.10: 87508265-87508265 on Build GRCh38
rs753858083 Chr.10: 87508269-87508269 on Build GRCh38
rs1044174458 Chr.10: 87508602-87508602 on Build GRCh38
rs766423567 Chr.10: 87508521-87508521 on Build GRCh38
rs140998185 Chr.10: 87508442-87508442 on Build GRCh38
rs1032900329 Chr.10: 87508653-87508653 on Build GRCh38
rs1035552815 Chr.10: 87508665-87508665 on Build GRCh38
rs141592439 Chr.10: 87508513-87508513 on Build GRCh38
rs761118032 Chr.10: 87508335-87508335 on Build GRCh38
rs765279000 Chr.10: 87508374-87508374 on Build GRCh38
rs751253859 Chr.10: 87508419-87508419 on Build GRCh38
rs767602733 Chr.10: 87508319-87508320 on Build GRCh38
rs752574762 Chr.10: 87508534-87508534 on Build GRCh38
rs779031027 Chr.10: 87508284-87508284 on Build GRCh38
rs146621374 Chr.10: 87508479-87508479 on Build GRCh38
rs369216745 Chr.10: 87508528-87508528 on Build GRCh38
rs891693838 Chr.10: 87508675-87508675 on Build GRCh38
rs9663404 Chr.10: 87508688-87508688 on Build GRCh38
rs762258613 Chr.10: 87508494-87508494 on Build GRCh38
rs575437529 Chr.10: 87508272-87508272 on Build GRCh38
rs754341204 Chr.10: 87508351-87508351 on Build GRCh38
rs1039266338 Chr.10: 87508369-87508369 on Build GRCh38
rs776720909 Chr.10: 87508318-87508318 on Build GRCh38
rs104894171 Chr.10: 87508507-87508507 on Build GRCh38
rs780770929 Chr.10: 87508339-87508339 on Build GRCh38
rs545242489 Chr.10: 87508455-87508455 on Build GRCh38
rs374832372 Chr.10: 87508421-87508421 on Build GRCh38
rs766090928 Chr.10: 87508329-87508329 on Build GRCh38
rs988563749 Chr.10: 87508670-87508670 on Build GRCh38
rs979445605 Chr.10: 87508729-87508729 on Build GRCh38
rs750537943 Chr.10: 87508387-87508387 on Build GRCh38
rs1010588044 Chr.10: 87508696-87508696 on Build GRCh38
rs781197534 Chr.10: 87508434-87508434 on Build GRCh38

View Full Product Details