Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113978048-113978545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASA3
Entrez Gene ID
22821
Gene Name
RAS p21 protein activator 3
Gene Aliases
GAP1IP4BP, GAPIII
UniGene
Hs.593075
Gene Chromosome Location
Chr.13: 113977783-114132623 on Build GRCh38
Transcripts
NM_001320821.1 NM_001320822.1 NM_007368.3 XM_011534841.2 XM_011534842.2 XM_017020438.1 XM_017020439.1

SNP Detail

SNP ID Chromosome Location
rs945216277 Chr.13: 113978465-113978465 on Build GRCh38
rs759476901 Chr.13: 113978084-113978084 on Build GRCh38
rs1054838976 Chr.13: 113978265-113978265 on Build GRCh38
rs1036543088 Chr.13: 113978167-113978167 on Build GRCh38
rs867119703 Chr.13: 113978472-113978472 on Build GRCh38
rs1027704619 Chr.13: 113978075-113978075 on Build GRCh38
rs959045120 Chr.13: 113978076-113978076 on Build GRCh38
rs974063634 Chr.13: 113978378-113978378 on Build GRCh38
rs944400708 Chr.13: 113978165-113978165 on Build GRCh38
rs931721739 Chr.13: 113978135-113978135 on Build GRCh38
rs569822464 Chr.13: 113978256-113978256 on Build GRCh38
rs998613112 Chr.13: 113978100-113978100 on Build GRCh38
rs1051340498 Chr.13: 113978108-113978108 on Build GRCh38
rs1000469156 Chr.13: 113978258-113978258 on Build GRCh38
rs565603008 Chr.13: 113978109-113978109 on Build GRCh38
rs903557259 Chr.13: 113978244-113978244 on Build GRCh38
rs557621017 Chr.13: 113978207-113978207 on Build GRCh38
rs971688472 Chr.13: 113978321-113978321 on Build GRCh38
rs556673144 Chr.13: 113978278-113978278 on Build GRCh38
rs971414340 Chr.13: 113978098-113978098 on Build GRCh38
rs11552973 Chr.13: 113978072-113978072 on Build GRCh38
rs118033094 Chr.13: 113978130-113978130 on Build GRCh38
rs1024716578 Chr.13: 113978097-113978097 on Build GRCh38
rs911535114 Chr.13: 113978161-113978161 on Build GRCh38
rs191023126 Chr.13: 113978348-113978348 on Build GRCh38
rs1042050113 Chr.13: 113978234-113978234 on Build GRCh38
rs921367869 Chr.13: 113978385-113978385 on Build GRCh38
rs548090196 Chr.13: 113978073-113978073 on Build GRCh38
rs182332294 Chr.13: 113978430-113978430 on Build GRCh38
rs1013403061 Chr.13: 113978233-113978233 on Build GRCh38
rs954232467 Chr.13: 113978387-113978387 on Build GRCh38
rs911661937 Chr.13: 113978510-113978510 on Build GRCh38
rs1030939510 Chr.13: 113978101-113978101 on Build GRCh38
rs112499290 Chr.13: 113978273-113978273 on Build GRCh38
rs867573636 Chr.13: 113978509-113978509 on Build GRCh38
rs145320115 Chr.13: 113978419-113978420 on Build GRCh38
rs930772522 Chr.13: 113978181-113978181 on Build GRCh38
rs898088959 Chr.13: 113978179-113978179 on Build GRCh38
rs998870592 Chr.13: 113978289-113978289 on Build GRCh38
rs572606093 Chr.13: 113978505-113978505 on Build GRCh38
rs1013041833 Chr.13: 113978077-113978077 on Build GRCh38
rs956614116 Chr.13: 113978352-113978352 on Build GRCh38
rs751643100 Chr.13: 113978151-113978151 on Build GRCh38
rs750801841 Chr.13: 113978288-113978288 on Build GRCh38
rs764280649 Chr.13: 113978131-113978131 on Build GRCh38
rs894674165 Chr.13: 113978208-113978208 on Build GRCh38
rs962745252 Chr.13: 113978356-113978356 on Build GRCh38
rs757453693 Chr.13: 113978191-113978191 on Build GRCh38
rs1042308637 Chr.13: 113978478-113978478 on Build GRCh38
rs912356370 Chr.13: 113978463-113978463 on Build GRCh38
rs756492080 Chr.13: 113978396-113978396 on Build GRCh38
rs1015838186 Chr.13: 113978345-113978345 on Build GRCh38
rs1004498854 Chr.13: 113978330-113978330 on Build GRCh38
rs894877866 Chr.13: 113978271-113978271 on Build GRCh38
rs974180592 Chr.13: 113978071-113978071 on Build GRCh38
rs542211822 Chr.13: 113978362-113978362 on Build GRCh38

View Full Product Details