Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49485661-49486243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
UniGene
Hs.567268
Gene Chromosome Location
Chr.15: 49423178-49487326 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

View Full Product Details