Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40556007-40556603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO1
Entrez Gene ID
2308
Gene Name
forkhead box O1
Gene Aliases
FKH1, FKHR, FOXO1A
UniGene
Hs.370666
Gene Chromosome Location
Chr.13: 40555664-40666597 on Build GRCh38
Transcripts
NM_002015.3 XM_011535008.2 XM_011535010.2

SNP Detail

SNP ID Chromosome Location
rs766433687 Chr.13: 40556205-40556205 on Build GRCh38
rs536362400 Chr.13: 40556151-40556151 on Build GRCh38
rs973272849 Chr.13: 40556579-40556579 on Build GRCh38
rs961866881 Chr.13: 40556520-40556520 on Build GRCh38
rs567201093 Chr.13: 40556083-40556083 on Build GRCh38
rs939358895 Chr.13: 40556391-40556391 on Build GRCh38
rs1014872161 Chr.13: 40556507-40556507 on Build GRCh38
rs568537590 Chr.13: 40556495-40556495 on Build GRCh38
rs907720873 Chr.13: 40556281-40556281 on Build GRCh38
rs1043974474 Chr.13: 40556071-40556071 on Build GRCh38
rs927993267 Chr.13: 40556380-40556380 on Build GRCh38
rs550249101 Chr.13: 40556465-40556465 on Build GRCh38
rs916625312 Chr.13: 40556464-40556464 on Build GRCh38
rs987294749 Chr.13: 40556270-40556270 on Build GRCh38
rs1031778671 Chr.13: 40556185-40556185 on Build GRCh38
rs188090269 Chr.13: 40556378-40556378 on Build GRCh38
rs193132822 Chr.13: 40556427-40556427 on Build GRCh38
rs372851785 Chr.13: 40556483-40556483 on Build GRCh38
rs940528830 Chr.13: 40556285-40556285 on Build GRCh38
rs1055626399 Chr.13: 40556392-40556392 on Build GRCh38
rs1043901510 Chr.13: 40556476-40556476 on Build GRCh38
rs566694615 Chr.13: 40556327-40556327 on Build GRCh38
rs536960434 Chr.13: 40556271-40556271 on Build GRCh38
rs998379920 Chr.13: 40556209-40556209 on Build GRCh38
rs148195072 Chr.13: 40556575-40556575 on Build GRCh38
rs973138152 Chr.13: 40556303-40556304 on Build GRCh38
rs920377434 Chr.13: 40556308-40556308 on Build GRCh38
rs1055211631 Chr.13: 40556048-40556048 on Build GRCh38
rs902745873 Chr.13: 40556090-40556090 on Build GRCh38
rs117958521 Chr.13: 40556536-40556536 on Build GRCh38
rs1036280547 Chr.13: 40556079-40556079 on Build GRCh38
rs1028097243 Chr.13: 40556522-40556522 on Build GRCh38
rs185321939 Chr.13: 40556531-40556531 on Build GRCh38
rs1045135853 Chr.13: 40556363-40556363 on Build GRCh38
rs1021690969 Chr.13: 40556199-40556199 on Build GRCh38
rs983929628 Chr.13: 40556264-40556264 on Build GRCh38
rs1010996325 Chr.13: 40556070-40556070 on Build GRCh38
rs895352138 Chr.13: 40556065-40556065 on Build GRCh38
rs756343090 Chr.13: 40556092-40556092 on Build GRCh38
rs999770958 Chr.13: 40556174-40556174 on Build GRCh38
rs889509461 Chr.13: 40556568-40556568 on Build GRCh38
rs953865158 Chr.13: 40556232-40556232 on Build GRCh38
rs1040622948 Chr.13: 40556487-40556487 on Build GRCh38
rs537048188 Chr.13: 40556512-40556512 on Build GRCh38
rs1010256872 Chr.13: 40556118-40556118 on Build GRCh38
rs546782697 Chr.13: 40556210-40556210 on Build GRCh38
rs891948868 Chr.13: 40556379-40556379 on Build GRCh38
rs900845354 Chr.13: 40556492-40556492 on Build GRCh38
rs904620236 Chr.13: 40556405-40556405 on Build GRCh38
rs902672447 Chr.13: 40556482-40556482 on Build GRCh38
rs961311930 Chr.13: 40556367-40556367 on Build GRCh38
rs947661784 Chr.13: 40556309-40556309 on Build GRCh38
rs35569244 Chr.13: 40556409-40556410 on Build GRCh38
rs796812663 Chr.13: 40556365-40556365 on Build GRCh38

View Full Product Details