Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87503777-87504287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MINPP1
Entrez Gene ID
9562
Gene Name
multiple inositol-polyphosphate phosphatase 1
Gene Aliases
HIPER1, MINPP2, MIPP
UniGene
Hs.121260
Gene Chromosome Location
Chr.10: 87502792-87553461 on Build GRCh38
Transcripts
XM_006718078.2 XM_017016965.1 NR_039825.1

Gene Symbol
MIR4678
Entrez Gene ID
100616296
Gene Name
microRNA 4678
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.10: 87503881-87503954 on Build GRCh38
Transcripts
XM_006718078.2 XM_017016965.1 NR_039825.1

SNP Detail

SNP ID Chromosome Location
rs757776719 Chr.10: 87504006-87504007 on Build GRCh38
rs561849290 Chr.10: 87504246-87504246 on Build GRCh38
rs571145407 Chr.10: 87504121-87504121 on Build GRCh38
rs746007280 Chr.10: 87504191-87504191 on Build GRCh38
rs549473510 Chr.10: 87503835-87503835 on Build GRCh38
rs990829604 Chr.10: 87503887-87503887 on Build GRCh38
rs932215331 Chr.10: 87504081-87504081 on Build GRCh38
rs1026921662 Chr.10: 87503822-87503822 on Build GRCh38
rs888344226 Chr.10: 87503832-87503832 on Build GRCh38
rs763488060 Chr.10: 87504140-87504140 on Build GRCh38
rs533551319 Chr.10: 87503954-87503954 on Build GRCh38
rs143602037 Chr.10: 87503819-87503819 on Build GRCh38
rs1001404820 Chr.10: 87503908-87503908 on Build GRCh38
rs867693359 Chr.10: 87504153-87504153 on Build GRCh38
rs549264056 Chr.10: 87504077-87504077 on Build GRCh38
rs556420197 Chr.10: 87504177-87504177 on Build GRCh38
rs1049520482 Chr.10: 87504160-87504160 on Build GRCh38
rs967616576 Chr.10: 87504149-87504149 on Build GRCh38
rs532893827 Chr.10: 87503909-87503909 on Build GRCh38
rs551176148 Chr.10: 87503936-87503936 on Build GRCh38
rs144044508 Chr.10: 87504259-87504259 on Build GRCh38
rs914825338 Chr.10: 87504145-87504145 on Build GRCh38
rs995508729 Chr.10: 87503973-87503973 on Build GRCh38
rs765537041 Chr.10: 87503944-87503944 on Build GRCh38
rs775190937 Chr.10: 87503833-87503833 on Build GRCh38
rs547751836 Chr.10: 87503952-87503952 on Build GRCh38
rs539609975 Chr.10: 87504206-87504206 on Build GRCh38
rs577896348 Chr.10: 87504193-87504193 on Build GRCh38
rs1048802964 Chr.10: 87503992-87503992 on Build GRCh38
rs921320687 Chr.10: 87504195-87504195 on Build GRCh38
rs978401775 Chr.10: 87503974-87503974 on Build GRCh38
rs1033408623 Chr.10: 87504084-87504084 on Build GRCh38
rs866097400 Chr.10: 87504152-87504152 on Build GRCh38
rs1048351409 Chr.10: 87504205-87504205 on Build GRCh38
rs562267419 Chr.10: 87503843-87503843 on Build GRCh38
rs970543038 Chr.10: 87504070-87504070 on Build GRCh38
rs74150470 Chr.10: 87504092-87504092 on Build GRCh38
rs957690459 Chr.10: 87503883-87503883 on Build GRCh38
rs1034233176 Chr.10: 87504183-87504183 on Build GRCh38
rs1004021756 Chr.10: 87503812-87503812 on Build GRCh38
rs989129631 Chr.10: 87504143-87504143 on Build GRCh38
rs959817371 Chr.10: 87503947-87503947 on Build GRCh38
rs897889242 Chr.10: 87504116-87504116 on Build GRCh38
rs2233161 Chr.10: 87504082-87504082 on Build GRCh38
rs931324221 Chr.10: 87504200-87504200 on Build GRCh38
rs887035037 Chr.10: 87504230-87504230 on Build GRCh38
rs550726521 Chr.10: 87503821-87503821 on Build GRCh38
rs888404409 Chr.10: 87504169-87504169 on Build GRCh38
rs573191100 Chr.10: 87504263-87504263 on Build GRCh38
rs926252247 Chr.10: 87504066-87504066 on Build GRCh38
rs887007373 Chr.10: 87504010-87504010 on Build GRCh38
rs1004458651 Chr.10: 87504053-87504053 on Build GRCh38
rs1036417460 Chr.10: 87504114-87504114 on Build GRCh38
rs967050672 Chr.10: 87503966-87503966 on Build GRCh38
rs912049476 Chr.10: 87504087-87504087 on Build GRCh38

View Full Product Details