Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 54695642-54695805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIT
Entrez Gene ID
3815
Gene Name
KIT proto-oncogene receptor tyrosine kinase
Gene Aliases
C-Kit, CD117, PBT, SCFR
UniGene
Hs.479754
Gene Chromosome Location
Chr.4: 54657928-54740715 on Build GRCh38
Transcripts
NM_000222.2 NM_001093772.1 XM_005265740.1 XM_005265741.1 XM_005265742.2 XM_017008178.1 XM_017008179.1 XM_017008180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6494994 c.252G>T p.T84T Chr.4: 54695696-54695696 on Build GRCh38
1430109 c.288G>A p.T96T Chr.4: 54695732-54695732 on Build GRCh38
6983718 c.291delC p.C97fs*1 Chr.4: 54695735-54695735 on Build GRCh38
1430110 c.303C>T p.H101H Chr.4: 54695747-54695747 on Build GRCh38
6437972 c.265G>A p.A89T Chr.4: 54695709-54695709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150026676 Chr.4: 54695732-54695732 on Build GRCh38
rs146081659 Chr.4: 54695724-54695724 on Build GRCh38
rs781130745 Chr.4: 54695745-54695745 on Build GRCh38
rs750916458 Chr.4: 54695687-54695687 on Build GRCh38
rs867439564 Chr.4: 54695730-54695730 on Build GRCh38
rs780922356 Chr.4: 54695764-54695764 on Build GRCh38
rs779113666 Chr.4: 54695729-54695729 on Build GRCh38
rs145333060 Chr.4: 54695747-54695747 on Build GRCh38
rs56411694 Chr.4: 54695696-54695696 on Build GRCh38
rs769632130 Chr.4: 54695748-54695748 on Build GRCh38
rs904550523 Chr.4: 54695725-54695725 on Build GRCh38
rs959320112 Chr.4: 54695738-54695738 on Build GRCh38
rs1020222899 Chr.4: 54695705-54695705 on Build GRCh38
rs371353189 Chr.4: 54695674-54695674 on Build GRCh38
rs768599276 Chr.4: 54695741-54695741 on Build GRCh38
rs373066995 Chr.4: 54695720-54695720 on Build GRCh38
rs779652404 Chr.4: 54695761-54695761 on Build GRCh38
rs749024557 Chr.4: 54695763-54695763 on Build GRCh38
rs794726674 Chr.4: 54695697-54695697 on Build GRCh38
rs201872586 Chr.4: 54695695-54695695 on Build GRCh38

View Full Product Details