Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 54698355-54698520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIT
Entrez Gene ID
3815
Gene Name
KIT proto-oncogene receptor tyrosine kinase
Gene Aliases
C-Kit, CD117, PBT, SCFR
UniGene
Hs.479754
Gene Chromosome Location
Chr.4: 54657928-54740715 on Build GRCh38
Transcripts
NM_000222.2 NM_001093772.1 XM_005265740.1 XM_005265741.1 XM_005265742.2 XM_017008178.1 XM_017008179.1 XM_017008180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6937118 c.490C>T p.P164S Chr.4: 54698436-54698436 on Build GRCh38
5609251 c.470C>T p.P157L Chr.4: 54698416-54698416 on Build GRCh38
6918874 c.409C>A p.P137T Chr.4: 54698355-54698355 on Build GRCh38
6696060 c.482G>T p.R161M Chr.4: 54698428-54698428 on Build GRCh38
5240903 c.530G>T p.R177L Chr.4: 54698476-54698476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774584792 Chr.4: 54698432-54698432 on Build GRCh38
rs200851152 Chr.4: 54698448-54698448 on Build GRCh38
rs145993517 Chr.4: 54698468-54698468 on Build GRCh38
rs115585711 Chr.4: 54698478-54698478 on Build GRCh38
rs775817289 Chr.4: 54698419-54698419 on Build GRCh38
rs777031731 Chr.4: 54698409-54698409 on Build GRCh38
rs532461931 Chr.4: 54698474-54698474 on Build GRCh38
rs764280416 Chr.4: 54698428-54698428 on Build GRCh38
rs367719489 Chr.4: 54698410-54698410 on Build GRCh38
rs777463519 Chr.4: 54698384-54698384 on Build GRCh38
rs145053429 Chr.4: 54698400-54698400 on Build GRCh38
rs767567085 Chr.4: 54698447-54698447 on Build GRCh38
rs758171174 Chr.4: 54698467-54698467 on Build GRCh38
rs746476450 Chr.4: 54698387-54698387 on Build GRCh38
rs758120380 Chr.4: 54698376-54698376 on Build GRCh38
rs776858609 Chr.4: 54698396-54698396 on Build GRCh38
rs763226471 Chr.4: 54698424-54698424 on Build GRCh38
rs544555667 Chr.4: 54698390-54698390 on Build GRCh38
rs752511532 Chr.4: 54698458-54698458 on Build GRCh38
rs770422212 Chr.4: 54698403-54698403 on Build GRCh38
rs878853767 Chr.4: 54698412-54698412 on Build GRCh38
rs201222895 Chr.4: 54698430-54698430 on Build GRCh38
rs140469176 Chr.4: 54698450-54698450 on Build GRCh38
rs770362669 Chr.4: 54698418-54698418 on Build GRCh38
rs149092990 Chr.4: 54698449-54698449 on Build GRCh38
rs746419269 Chr.4: 54698489-54698489 on Build GRCh38
rs1003144287 Chr.4: 54698380-54698380 on Build GRCh38
rs761831851 Chr.4: 54698444-54698444 on Build GRCh38
rs756722358 Chr.4: 54698477-54698477 on Build GRCh38

View Full Product Details