Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40559666-40559839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO1
Entrez Gene ID
2308
Gene Name
forkhead box O1
Gene Aliases
FKH1, FKHR, FOXO1A
UniGene
Hs.370666
Gene Chromosome Location
Chr.13: 40555664-40666597 on Build GRCh38
Transcripts
NM_002015.3 XM_011535008.2 XM_011535010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947358 c.1763G>A p.G588D Chr.13: 40559728-40559728 on Build GRCh38
374141 c.1770G>T p.M590I Chr.13: 40559721-40559721 on Build GRCh38
5601523 c.1779C>T p.L593L Chr.13: 40559712-40559712 on Build GRCh38
1207452 c.1721C>A p.A574D Chr.13: 40559770-40559770 on Build GRCh38
1940589 c.1739C>T p.S580F Chr.13: 40559752-40559752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373830610 Chr.13: 40559785-40559785 on Build GRCh38
rs775906686 Chr.13: 40559712-40559712 on Build GRCh38
rs201186715 Chr.13: 40559803-40559803 on Build GRCh38
rs767915535 Chr.13: 40559734-40559734 on Build GRCh38
rs540866852 Chr.13: 40559714-40559714 on Build GRCh38
rs372159853 Chr.13: 40559709-40559709 on Build GRCh38
rs150896957 Chr.13: 40559755-40559755 on Build GRCh38
rs757585400 Chr.13: 40559731-40559731 on Build GRCh38
rs751994432 Chr.13: 40559728-40559728 on Build GRCh38
rs867332663 Chr.13: 40559699-40559699 on Build GRCh38
rs747905007 Chr.13: 40559758-40559758 on Build GRCh38
rs773141065 Chr.13: 40559700-40559700 on Build GRCh38
rs775747228 Chr.13: 40559807-40559807 on Build GRCh38
rs773087473 Chr.13: 40559777-40559777 on Build GRCh38
rs950234021 Chr.13: 40559788-40559788 on Build GRCh38
rs148727582 Chr.13: 40559735-40559735 on Build GRCh38
rs746945626 Chr.13: 40559792-40559792 on Build GRCh38
rs564402015 Chr.13: 40559715-40559715 on Build GRCh38
rs755827604 Chr.13: 40559750-40559750 on Build GRCh38
rs750150806 Chr.13: 40559710-40559710 on Build GRCh38
rs770037212 Chr.13: 40559805-40559805 on Build GRCh38
rs377056137 Chr.13: 40559751-40559751 on Build GRCh38
rs45491494 Chr.13: 40559754-40559754 on Build GRCh38
rs906022358 Chr.13: 40559810-40559810 on Build GRCh38
rs771830343 Chr.13: 40559761-40559761 on Build GRCh38

View Full Product Details