Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 54703609-54703763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIT
Entrez Gene ID
3815
Gene Name
KIT proto-oncogene receptor tyrosine kinase
Gene Aliases
C-Kit, CD117, PBT, SCFR
UniGene
Hs.479754
Gene Chromosome Location
Chr.4: 54657928-54740715 on Build GRCh38
Transcripts
NM_000222.2 NM_001093772.1 XM_005265740.1 XM_005265741.1 XM_005265742.2 XM_017008178.1 XM_017008179.1 XM_017008180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5945005 c.794G>A p.G265D Chr.4: 54703761-54703761 on Build GRCh38
7201133 c.767A>T p.Q256L Chr.4: 54703734-54703734 on Build GRCh38
6970309 c.782G>T p.S261I Chr.4: 54703749-54703749 on Build GRCh38
3301432 c.793G>A p.G265S Chr.4: 54703760-54703760 on Build GRCh38
5356246 c.793G>T p.G265C Chr.4: 54703760-54703760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018986192 Chr.4: 54703654-54703654 on Build GRCh38
rs367986084 Chr.4: 54703715-54703715 on Build GRCh38
rs989062521 Chr.4: 54703693-54703693 on Build GRCh38
rs377724954 Chr.4: 54703682-54703682 on Build GRCh38
rs771144014 Chr.4: 54703680-54703680 on Build GRCh38
rs768605848 Chr.4: 54703688-54703688 on Build GRCh38
rs747157762 Chr.4: 54703675-54703675 on Build GRCh38
rs774060921 Chr.4: 54703692-54703692 on Build GRCh38
rs760308110 Chr.4: 54703717-54703717 on Build GRCh38
rs765944197 Chr.4: 54703724-54703724 on Build GRCh38
rs927816551 Chr.4: 54703666-54703666 on Build GRCh38
rs768559669 Chr.4: 54703694-54703694 on Build GRCh38
rs544780604 Chr.4: 54703693-54703694 on Build GRCh38
rs530896729 Chr.4: 54703703-54703703 on Build GRCh38
rs926098687 Chr.4: 54703691-54703691 on Build GRCh38
rs550978864 Chr.4: 54703706-54703706 on Build GRCh38
rs571756237 Chr.4: 54703702-54703702 on Build GRCh38

View Full Product Details