Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92962262-92962495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts
XM_017021262.1 XM_017021263.1 XM_017021264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7127160 c.502G>A p.E168K Chr.14: 92962357-92962357 on Build GRCh38
6688585 c.488G>A p.G163D Chr.14: 92962371-92962371 on Build GRCh38
6688584 c.488G>A p.G163D Chr.14: 92962371-92962371 on Build GRCh38
7127161 c.502G>A p.E168K Chr.14: 92962357-92962357 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749221560 Chr.14: 92962365-92962365 on Build GRCh38
rs766654926 Chr.14: 92962289-92962289 on Build GRCh38
rs1044640241 Chr.14: 92962396-92962396 on Build GRCh38
rs369612289 Chr.14: 92962406-92962406 on Build GRCh38
rs780134021 Chr.14: 92962358-92962358 on Build GRCh38
rs894811335 Chr.14: 92962326-92962326 on Build GRCh38
rs2295393 Chr.14: 92962457-92962457 on Build GRCh38
rs754940470 Chr.14: 92962280-92962280 on Build GRCh38
rs772648130 Chr.14: 92962400-92962400 on Build GRCh38
rs777933712 Chr.14: 92962321-92962321 on Build GRCh38
rs371949086 Chr.14: 92962317-92962317 on Build GRCh38
rs1030222375 Chr.14: 92962469-92962469 on Build GRCh38
rs1031295219 Chr.14: 92962327-92962327 on Build GRCh38
rs923652052 Chr.14: 92962300-92962300 on Build GRCh38
rs765917398 Chr.14: 92962428-92962428 on Build GRCh38
rs771712816 Chr.14: 92962386-92962386 on Build GRCh38
rs765600617 Chr.14: 92962446-92962446 on Build GRCh38
rs569645568 Chr.14: 92962422-92962422 on Build GRCh38
rs999216197 Chr.14: 92962318-92962318 on Build GRCh38
rs910309032 Chr.14: 92962440-92962440 on Build GRCh38
rs747100670 Chr.14: 92962328-92962328 on Build GRCh38
rs996590883 Chr.14: 92962467-92962467 on Build GRCh38
rs538225581 Chr.14: 92962439-92962439 on Build GRCh38
rs957269790 Chr.14: 92962369-92962369 on Build GRCh38
rs771194929 Chr.14: 92962352-92962352 on Build GRCh38
rs768475112 Chr.14: 92962368-92962368 on Build GRCh38
rs7161091 Chr.14: 92962466-92962466 on Build GRCh38
rs1012736536 Chr.14: 92962297-92962297 on Build GRCh38
rs759987080 Chr.14: 92962442-92962442 on Build GRCh38
rs1003019826 Chr.14: 92962423-92962423 on Build GRCh38
rs886640740 Chr.14: 92962445-92962445 on Build GRCh38
rs558428114 Chr.14: 92962455-92962455 on Build GRCh38
rs774037199 Chr.14: 92962379-92962379 on Build GRCh38
rs761482275 Chr.14: 92962385-92962385 on Build GRCh38
rs929522478 Chr.14: 92962463-92962463 on Build GRCh38
rs976207194 Chr.14: 92962444-92962444 on Build GRCh38
rs377621405 Chr.14: 92962296-92962296 on Build GRCh38
rs540513924 Chr.14: 92962459-92962459 on Build GRCh38
rs777948550 Chr.14: 92962408-92962408 on Build GRCh38

View Full Product Details