Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49424363-49424620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Gene Symbol
FGF7
Entrez Gene ID
2252
Gene Name
fibroblast growth factor 7
Gene Aliases
HBGF-7, KGF
UniGene
Hs.567268
Gene Chromosome Location
Chr.15: 49423178-49487326 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_011521324.1 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_002009.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
962478 c.131T>C p.V44A Chr.15: 49424428-49424428 on Build GRCh38
962479 c.161G>T p.R54I Chr.15: 49424458-49424458 on Build GRCh38
5722313 c.70C>T p.L24L Chr.15: 49424367-49424367 on Build GRCh38
4055403 c.238G>A p.D80N Chr.15: 49424535-49424535 on Build GRCh38
433998 c.115C>T p.Q39* Chr.15: 49424412-49424412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746255850 Chr.15: 49424400-49424400 on Build GRCh38
rs746522643 Chr.15: 49424432-49424432 on Build GRCh38
rs765581354 Chr.15: 49424591-49424592 on Build GRCh38
rs768087610 Chr.15: 49424438-49424438 on Build GRCh38
rs758913734 Chr.15: 49424404-49424404 on Build GRCh38
rs747759102 Chr.15: 49424483-49424483 on Build GRCh38
rs1042215908 Chr.15: 49424527-49424527 on Build GRCh38
rs779375910 Chr.15: 49424398-49424398 on Build GRCh38
rs753997634 Chr.15: 49424552-49424552 on Build GRCh38
rs764225350 Chr.15: 49424550-49424550 on Build GRCh38
rs769477922 Chr.15: 49424486-49424486 on Build GRCh38
rs745689304 Chr.15: 49424489-49424489 on Build GRCh38
rs370236693 Chr.15: 49424502-49424502 on Build GRCh38
rs34531231 Chr.15: 49424473-49424473 on Build GRCh38
rs1057133325 Chr.15: 49424414-49424414 on Build GRCh38
rs376909937 Chr.15: 49424418-49424418 on Build GRCh38
rs895834784 Chr.15: 49424422-49424422 on Build GRCh38
rs776131808 Chr.15: 49424472-49424472 on Build GRCh38
rs762071927 Chr.15: 49424584-49424584 on Build GRCh38
rs774091678 Chr.15: 49424537-49424537 on Build GRCh38
rs529145599 Chr.15: 49424504-49424504 on Build GRCh38
rs766338451 Chr.15: 49424511-49424511 on Build GRCh38
rs143883386 Chr.15: 49424542-49424542 on Build GRCh38
rs762092181 Chr.15: 49424581-49424581 on Build GRCh38
rs1010311644 Chr.15: 49424468-49424468 on Build GRCh38
rs17850705 Chr.15: 49424482-49424482 on Build GRCh38
rs375963598 Chr.15: 49424586-49424586 on Build GRCh38
rs183624878 Chr.15: 49424519-49424519 on Build GRCh38
rs780482125 Chr.15: 49424420-49424420 on Build GRCh38

View Full Product Details