Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92962810-92963035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts
XM_017021262.1 XM_017021263.1 XM_017021264.1

SNP Detail

SNP ID Chromosome Location
rs769016989 Chr.14: 92962971-92962971 on Build GRCh38
rs778700158 Chr.14: 92962833-92962833 on Build GRCh38
rs202143789 Chr.14: 92962878-92962878 on Build GRCh38
rs542826472 Chr.14: 92962969-92962969 on Build GRCh38
rs916309062 Chr.14: 92962834-92962834 on Build GRCh38
rs969547602 Chr.14: 92962845-92962845 on Build GRCh38
rs371550689 Chr.14: 92963006-92963007 on Build GRCh38
rs35134553 Chr.14: 92962937-92962937 on Build GRCh38
rs748180262 Chr.14: 92962841-92962841 on Build GRCh38
rs758366986 Chr.14: 92962907-92962907 on Build GRCh38
rs1051896575 Chr.14: 92962978-92962978 on Build GRCh38
rs369880309 Chr.14: 92962893-92962893 on Build GRCh38
rs999513316 Chr.14: 92963004-92963004 on Build GRCh38
rs770578480 Chr.14: 92962861-92962861 on Build GRCh38
rs746629240 Chr.14: 92962848-92962848 on Build GRCh38
rs780560878 Chr.14: 92962906-92962906 on Build GRCh38
rs932050347 Chr.14: 92962958-92962958 on Build GRCh38
rs574899865 Chr.14: 92962865-92962865 on Build GRCh38
rs890438629 Chr.14: 92962991-92962991 on Build GRCh38
rs754882912 Chr.14: 92962829-92962829 on Build GRCh38
rs183824704 Chr.14: 92962866-92962866 on Build GRCh38
rs745645901 Chr.14: 92962894-92962894 on Build GRCh38
rs774886170 Chr.14: 92962880-92962880 on Build GRCh38
rs554028633 Chr.14: 92962889-92962889 on Build GRCh38
rs749847947 Chr.14: 92962921-92962921 on Build GRCh38
rs1030848044 Chr.14: 92963006-92963006 on Build GRCh38
rs776150632 Chr.14: 92962864-92962864 on Build GRCh38
rs989732881 Chr.14: 92962830-92962830 on Build GRCh38
rs558308297 Chr.14: 92962847-92962847 on Build GRCh38
rs114560172 Chr.14: 92962922-92962922 on Build GRCh38
rs775830614 Chr.14: 92962869-92962869 on Build GRCh38
rs756140902 Chr.14: 92962900-92962900 on Build GRCh38
rs763601682 Chr.14: 92962877-92962877 on Build GRCh38
rs150264238 Chr.14: 92962846-92962846 on Build GRCh38
rs762057168 Chr.14: 92962881-92962881 on Build GRCh38
rs1016466976 Chr.14: 92962968-92962968 on Build GRCh38

View Full Product Details