Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27362352-27362619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_005255308.2 XM_006721043.1 XM_011545825.1 XM_011545826.2 XM_011545827.2 XM_011545828.2 XM_011545829.1 XM_011545830.2 XM_011545831.1 XM_011545832.1 XM_011545833.2 XM_011545834.2 XM_017023211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3794727 c.1252G>A p.E418K Chr.16: 27362604-27362604 on Build GRCh38
7372118 c.1259G>T p.G420V Chr.16: 27362611-27362611 on Build GRCh38
3402220 c.1072G>A p.V358M Chr.16: 27362424-27362424 on Build GRCh38
3772176 c.1079G>A p.R360Q Chr.16: 27362431-27362431 on Build GRCh38
148058 c.1199A>C p.E400A Chr.16: 27362551-27362551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978321639 Chr.16: 27362395-27362395 on Build GRCh38
rs751339888 Chr.16: 27362434-27362434 on Build GRCh38
rs142898853 Chr.16: 27362403-27362403 on Build GRCh38
rs915680769 Chr.16: 27362436-27362436 on Build GRCh38
rs141632606 Chr.16: 27362452-27362452 on Build GRCh38
rs762906703 Chr.16: 27362522-27362522 on Build GRCh38
rs778127963 Chr.16: 27362430-27362430 on Build GRCh38
rs772745695 Chr.16: 27362571-27362571 on Build GRCh38
rs767364258 Chr.16: 27362599-27362599 on Build GRCh38
rs372264723 Chr.16: 27362424-27362424 on Build GRCh38
rs767857685 Chr.16: 27362391-27362391 on Build GRCh38
rs775887007 Chr.16: 27362482-27362482 on Build GRCh38
rs748338395 Chr.16: 27362377-27362377 on Build GRCh38
rs746330200 Chr.16: 27362456-27362456 on Build GRCh38
rs769465913 Chr.16: 27362567-27362567 on Build GRCh38
rs776329120 Chr.16: 27362498-27362498 on Build GRCh38
rs796971054 Chr.16: 27362578-27362578 on Build GRCh38
rs376032165 Chr.16: 27362565-27362565 on Build GRCh38
rs558584235 Chr.16: 27362402-27362402 on Build GRCh38
rs35357296 Chr.16: 27362597-27362597 on Build GRCh38
rs760989912 Chr.16: 27362486-27362486 on Build GRCh38
rs146226201 Chr.16: 27362496-27362496 on Build GRCh38
rs1805011 Chr.16: 27362551-27362551 on Build GRCh38
rs6413500 Chr.16: 27362512-27362512 on Build GRCh38
rs775057596 Chr.16: 27362372-27362372 on Build GRCh38
rs200547374 Chr.16: 27362433-27362433 on Build GRCh38
rs759816754 Chr.16: 27362390-27362390 on Build GRCh38
rs766265918 Chr.16: 27362528-27362528 on Build GRCh38
rs768938613 Chr.16: 27362563-27362563 on Build GRCh38
rs761456239 Chr.16: 27362510-27362510 on Build GRCh38
rs552217920 Chr.16: 27362562-27362562 on Build GRCh38
rs139061387 Chr.16: 27362431-27362431 on Build GRCh38
rs889955602 Chr.16: 27362546-27362546 on Build GRCh38
rs749920780 Chr.16: 27362513-27362513 on Build GRCh38
rs200733686 Chr.16: 27362566-27362566 on Build GRCh38
rs772482594 Chr.16: 27362481-27362481 on Build GRCh38
rs150787891 Chr.16: 27362405-27362405 on Build GRCh38
rs778188392 Chr.16: 27362450-27362450 on Build GRCh38
rs375367781 Chr.16: 27362455-27362455 on Build GRCh38
rs200110537 Chr.16: 27362540-27362540 on Build GRCh38
rs754048279 Chr.16: 27362544-27362544 on Build GRCh38
rs747391679 Chr.16: 27362557-27362557 on Build GRCh38
rs2234898 Chr.16: 27362594-27362594 on Build GRCh38
rs751345478 Chr.16: 27362536-27362536 on Build GRCh38
rs951922180 Chr.16: 27362425-27362425 on Build GRCh38
rs763770991 Chr.16: 27362399-27362399 on Build GRCh38
rs759552353 Chr.16: 27362598-27362598 on Build GRCh38
rs757568489 Chr.16: 27362545-27362545 on Build GRCh38
rs756328536 Chr.16: 27362438-27362438 on Build GRCh38
rs756906772 Chr.16: 27362465-27362466 on Build GRCh38
rs762418643 Chr.16: 27362576-27362576 on Build GRCh38
rs994192168 Chr.16: 27362554-27362554 on Build GRCh38
rs774578296 Chr.16: 27362466-27362468 on Build GRCh38
rs775734293 Chr.16: 27362393-27362393 on Build GRCh38
rs754878179 Chr.16: 27362537-27362537 on Build GRCh38
rs146853841 Chr.16: 27362552-27362552 on Build GRCh38
rs538648103 Chr.16: 27362396-27362396 on Build GRCh38

View Full Product Details