Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109943639-109943912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGF
Entrez Gene ID
1950
Gene Name
epidermal growth factor
Gene Aliases
HOMG4, URG
UniGene
Hs.419815
Gene Chromosome Location
Chr.4: 109912883-110013079 on Build GRCh38
Transcripts
NM_001178130.2 NM_001178131.2 NM_001963.5 XM_005262796.2 XM_011531707.1 XM_017007845.1 XM_017007846.1 XM_017007847.1 XM_017007848.1 XM_017007849.1 XM_017007850.1 XM_017007851.1 XM_017007852.1 XM_017007853.1 XM_017007854.1 XM_017007855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4552178 c.551G>A p.R184K Chr.4: 109943883-109943883 on Build GRCh38
3599102 c.531G>A p.E177E Chr.4: 109943863-109943863 on Build GRCh38
6437729 c.522G>C p.W174C Chr.4: 109943854-109943854 on Build GRCh38
1050012 c.518T>G p.F173C Chr.4: 109943850-109943850 on Build GRCh38
5499509 c.562G>A p.D188N Chr.4: 109943894-109943894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150908894 Chr.4: 109943674-109943675 on Build GRCh38
rs544835493 Chr.4: 109943773-109943773 on Build GRCh38
rs1005569755 Chr.4: 109943808-109943808 on Build GRCh38
rs768345018 Chr.4: 109943810-109943810 on Build GRCh38
rs942069404 Chr.4: 109943783-109943783 on Build GRCh38
rs774628592 Chr.4: 109943879-109943879 on Build GRCh38
rs11568900 Chr.4: 109943782-109943782 on Build GRCh38
rs1039066470 Chr.4: 109943795-109943795 on Build GRCh38
rs138711485 Chr.4: 109943893-109943893 on Build GRCh38
rs28553469 Chr.4: 109943888-109943888 on Build GRCh38
rs544300403 Chr.4: 109943830-109943830 on Build GRCh38
rs553251458 Chr.4: 109943760-109943760 on Build GRCh38
rs952574092 Chr.4: 109943803-109943803 on Build GRCh38
rs530568156 Chr.4: 109943821-109943821 on Build GRCh38
rs572677232 Chr.4: 109943762-109943762 on Build GRCh38
rs542523380 Chr.4: 109943730-109943730 on Build GRCh38
rs369090159 Chr.4: 109943884-109943884 on Build GRCh38
rs11098055 Chr.4: 109943675-109943675 on Build GRCh38
rs573988680 Chr.4: 109943723-109943723 on Build GRCh38
rs895624761 Chr.4: 109943754-109943754 on Build GRCh38
rs747964253 Chr.4: 109943819-109943819 on Build GRCh38
rs375101300 Chr.4: 109943828-109943828 on Build GRCh38
rs376540740 Chr.4: 109943675-109943676 on Build GRCh38
rs776232830 Chr.4: 109943813-109943813 on Build GRCh38
rs937676505 Chr.4: 109943684-109943684 on Build GRCh38
rs905631090 Chr.4: 109943690-109943690 on Build GRCh38
rs28684716 Chr.4: 109943702-109943702 on Build GRCh38
rs1047891468 Chr.4: 109943781-109943781 on Build GRCh38

View Full Product Details